DNA Polymerase I, Large (Klenow) Fragment is a DNA polymerase enzyme that lacks the 5' to 3' exonuclease activity of intact DNA Polymerase I, but does exhibit the 5' to 3' DNA polymerase and 3' to 5' exonuclease activities.
Applications: Fill-in of 5´ overhangs (1). Synthesis of probes by random primers labeling method (2). Sequencing single- and double-stranded DNA (3). Site-directed mutagenesis.
Source: Purified from E. coli expressing the Klenow fragment on a plasmid.
Performance and Quality Testing: SDS-PAGE purity; single-stranded and double-stranded endodeoxyribonuclease and self-priming assays; performance evaluated in fill-in reaction.
Unit Definition: One unit incorporates 10 nmol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using a template•primer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description
DNA Polymerase
Polymerase
DNA Polymerase I
Exonuclease Activity
3' to 5'
Quantity
500 Units
Shipping Condition
Approved for shipment on Wet or Dry Ice
Hot Start
No
Contents & Storage
DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500 mM Tris-HCl (pH 8.0), 100 mM MgCl2 , 500 mM NaCl], vial of dilution buffer. Store at -20°C.