Boric acid, 99+%, extra pure
Boric acid, 99+%, extra pure
Boric acid, 99+%, extra pure
Boric acid, 99+%, extra pure
Thermo Scientific Chemicals

Boric acid, 99+%, extra pure

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
10 kg
1 kg
Packaging:
View all
Plastic bottle
Catalog number 180570025
Price (USD)
112.65
Special offer
Online exclusive
Ends: 26-Sep-2025
139.00
Save 26.35 (19%)
Each
In stock
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
112.65
Special offer
Online exclusive
Ends: 26-Sep-2025
139.00
Save 26.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystals or granules
Titration with NaOH99.0 to 101.0 %
Chloride (Cl)=<10 ppm
Iron (Fe)=<10 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473173Certificate of AnalysisJun 05, 2025180570100, 180570025, 11904101, 180570010
2538592Certificate of AnalysisMay 24, 2025180570100, 180570025, 11904101, 180570010
2535090Certificate of AnalysisMay 24, 2025180570100, 180570025, 11904101, 180570010
A0469339Certificate of AnalysisFeb 14, 2025180570100, 180570025, 180570010
2182543Certificate of AnalysisJan 27, 2025180570100, 180570025, 180570010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection