Thermo Scientific™

Orion™ 2100 Series and 1800 Series Analyzer Calibration Standards for Orion™ Online Monitors

Catalog number: 181040
Have Questions?
Thermo Scientific™

Orion™ 2100 Series and 1800 Series Analyzer Calibration Standards for Orion™ Online Monitors

Catalog number: 181040
Have Questions?
Use Thermo Scientific™ Orion™ 2100 Series and 1800 Series Analyzer Calibration Standards for Orion™ Online Monitors with Thermo Scientific™ Orion™ process analyzers.
 
Catalog Number
181040
Unit Size
Each
Description
Ammonia 191and 1920ppm Standards for 2110XP Ammonia Analyzers, 2 x 60mL Each
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionAmmonia 191and 1920ppm Standards for 2110XP Ammonia Analyzers, 2 x 60mL Each
TypeStandards
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Fluoride 1000ppm Standard for 2109XP Fluoride Analyzers, 475mL
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionFluoride 1000ppm Standard for 2109XP Fluoride Analyzers, 475mL
TypeStandards
For Use With (Equipment)Thermo Scientific™ 2100 Series Fluoride Analyzer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Hydrazine 100ppm Standard for 2118XP Oxygen Scavenger Analyzers, 475mL
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionHydrazine 100ppm Standard for 2118XP Oxygen Scavenger Analyzers, 475mL
TypeStandards
For Use With (Equipment)Thermo Scientific™ 2100 Series Oxygen Scavenger Analyzer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Fluoride 100and 1000ppm Standards for 2109XP Fluoride Analyzers, 2 x 60mL Each
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionFluoride 100and 1000ppm Standards for 2109XP Fluoride Analyzers, 2 x 60mL Each
TypeStandards
For Use With (Equipment)Thermo Scientific™ 2100 Series Fluoride Analyzer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sodium Electrode Etch Solution for 2111LL and 2111XP Sodium Analyzers, 30 mL
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSodium Electrode Etch Solution for 2111LL and 2111XP Sodium Analyzers, 30 mL
TypeEtch Solution
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sodium 19.1and 192ppm Standards for 2111LL and 2111XP Sodium Analyzers, 2 x 60 mL Each with Sodium Electrode Etch Solution
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSodium 19.1and 192ppm Standards for 2111LL and 2111XP Sodium Analyzers, 2 x 60 mL Each with Sodium Electrode Etch Solution
TypeStandards
For Use With (Equipment)Thermo Scientific™ 2100 Series Sodium Analyzer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sodium 1910and 19200ppm Standards for 2111LL and 2111XP Sodium Analyzers, 2 x 60 mL Each with Sodium Electrode Etch Solution
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSodium 1910and 19200ppm Standards for 2111LL and 2111XP Sodium Analyzers, 2 x 60 mL Each with Sodium Electrode Etch Solution
TypeStandards
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Calcium 191and 1920ppm Standards for 2120XP Calcium Hardness Analyzers, 2 x 60mL Each
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionCalcium 191and 1920ppm Standards for 2120XP Calcium Hardness Analyzers, 2 x 60mL Each
TypeStandards
Unit SizeEach
Catalog Number
Unit Size
Each
Description
pH Consumables Kit for 2100 Series pH Analyzers, 475 mL Each pH 4, 7 and 10 Buffers and Process ROSS Electrode Fill Solution
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionpH Consumables Kit for 2100 Series pH Analyzers, 475 mL Each pH 4, 7 and 10 Buffers and Process ROSS Electrode Fill Solution
TypeStandards
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Chloride 296and 2946ppm Standards for 2117XP Chloride Analyzers, 2 x 60mL Each
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionChloride 296and 2946ppm Standards for 2117XP Chloride Analyzers, 2 x 60mL Each
TypeStandards
Unit SizeEach
Showing 10 of 11Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
181040Full specifications
EachAmmonia 191and 1920ppm Standards for 2110XP Ammonia Analyzers, 2 x 60mL Each
-
DescriptionAmmonia 191and 1920ppm Standards for 2110XP Ammonia Analyzers, 2 x 60mL Each
TypeStandards
Unit SizeEach
110902Full specifications
EachFluoride 1000ppm Standard for 2109XP Fluoride Analyzers, 475mL
-
DescriptionFluoride 1000ppm Standard for 2109XP Fluoride Analyzers, 475mL
TypeStandards
For Use With (Equipment)Thermo Scientific™ 2100 Series Fluoride Analyzer
Unit SizeEach
151810Full specifications
EachHydrazine 100ppm Standard for 2118XP Oxygen Scavenger Analyzers, 475mL
-
DescriptionHydrazine 100ppm Standard for 2118XP Oxygen Scavenger Analyzers, 475mL
TypeStandards
For Use With (Equipment)Thermo Scientific™ 2100 Series Oxygen Scavenger Analyzer
Unit SizeEach
180940Full specifications
EachFluoride 100and 1000ppm Standards for 2109XP Fluoride Analyzers, 2 x 60mL Each
-
DescriptionFluoride 100and 1000ppm Standards for 2109XP Fluoride Analyzers, 2 x 60mL Each
TypeStandards
For Use With (Equipment)Thermo Scientific™ 2100 Series Fluoride Analyzer
Unit SizeEach
181113Full specifications
EachSodium Electrode Etch Solution for 2111LL and 2111XP Sodium Analyzers, 30 mL
-
DescriptionSodium Electrode Etch Solution for 2111LL and 2111XP Sodium Analyzers, 30 mL
TypeEtch Solution
Unit SizeEach
181140Full specifications
EachSodium 19.1and 192ppm Standards for 2111LL and 2111XP Sodium Analyzers, 2 x 60 mL Each with Sodium Electrode Etch Solution
-
DescriptionSodium 19.1and 192ppm Standards for 2111LL and 2111XP Sodium Analyzers, 2 x 60 mL Each with Sodium Electrode Etch Solution
TypeStandards
For Use With (Equipment)Thermo Scientific™ 2100 Series Sodium Analyzer
Unit SizeEach
181141Full specifications
EachSodium 1910and 19200ppm Standards for 2111LL and 2111XP Sodium Analyzers, 2 x 60 mL Each with Sodium Electrode Etch Solution
-
DescriptionSodium 1910and 19200ppm Standards for 2111LL and 2111XP Sodium Analyzers, 2 x 60 mL Each with Sodium Electrode Etch Solution
TypeStandards
Unit SizeEach
182040Full specifications
EachCalcium 191and 1920ppm Standards for 2120XP Calcium Hardness Analyzers, 2 x 60mL Each
-
DescriptionCalcium 191and 1920ppm Standards for 2120XP Calcium Hardness Analyzers, 2 x 60mL Each
TypeStandards
Unit SizeEach
2001CPFull specifications
EachpH Consumables Kit for 2100 Series pH Analyzers, 475 mL Each pH 4, 7 and 10 Buffers and Process ROSS Electrode Fill Solution
-
DescriptionpH Consumables Kit for 2100 Series pH Analyzers, 475 mL Each pH 4, 7 and 10 Buffers and Process ROSS Electrode Fill Solution
TypeStandards
Unit SizeEach
211740Full specifications
EachChloride 296and 2946ppm Standards for 2117XP Chloride Analyzers, 2 x 60mL Each
-
DescriptionChloride 296and 2946ppm Standards for 2117XP Chloride Analyzers, 2 x 60mL Each
TypeStandards
Unit SizeEach
Showing 10 of 11Show more
Calibration standards and solutions for the Orion 2109XP, 2110XP, 2111LL, 2111XP, 2117HL, 2117XP, 2118XP, 2120XP, 2100 pH, 2001 pH, 1809, 1810, 1811AO, 1811EL, 1817HL, 1818 and 1820 analyzers.

Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text