Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
Reduces downtime and labor spent on decontamination
Specifications
DescriptionInternal chamber replacement kit with HEPA, gaskets, and other parts
For Use With (Equipment)For Water Jacket CO2 Incubators
LightingLED
TypeDecontamination Kit
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.