Search Thermo Fisher Scientific
Use the Thermo Scientific™ iConnect™ Cold-On-Column Injector Module to benefit from true cold injection and ensure no thermal breakdown, no sample discrimination, and 100% recovery of heavier compounds.
Ensure 100% recovery during injection of high boiling compounds with the Thermo Scientific™ instant connect (iConnect™) Cold-On-Column (COC) Injector Module. When exposed to GC oven temperatures, direct deposition of liquid samples into a GC column with this injector negates the need for sample vaporization and transfer at high temperatures prior to separation, eliminating discrimination effects. This injector module is compatible with both the TRACE™ 1300 and 1600 Series Gas Chromatographs. Use the iConnect COC injector for analysis of high boiling compounds like triglycerides, heavy hydrocarbons, and waxes or thermolabile compounds sensitive to thermal breakdown during vaporization.
Reduce the risk of discrimination for high boiling compounds and assure quantitiative recovery of your sample.
Raise your gas chromatography analysis productivity and reduce cost of ownership with our customizable GC solutions, designed to fit any laboratory workflow.
Share catalog number, name or link