Cobalt(II) chloride hexahydrate, pure
Cobalt(II) chloride hexahydrate, pure
Cobalt(II) chloride hexahydrate, pure
Cobalt(II) chloride hexahydrate, pure
Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, pure

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 192862500
Price (USD)
80.65
Special offer
Online exclusive
94.90
Save 14.25 (15%)
Each
In stock
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
94.90
Save 14.25 (15%)
Each
Add to cart
Chemical Identifiers
CAS7791-13-1
IUPAC Nameλ²-cobalt(2+) hexahydrate dichloride
Molecular FormulaCl2CoH12O6
InChI KeyGFHNAMRJFCEERV-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to violet
Appearance (Form)Crystals
Titration Complexometric98.0 to 102.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471169Certificate of AnalysisMay 17, 2025192860010, 192862500
A0468763Certificate of AnalysisJan 25, 2025192860010, 192862500
A0465255Certificate of AnalysisAug 21, 2024192860010, 192862500
A0459435Certificate of AnalysisJan 23, 2024192860010, 192862500
A0456939Certificate of AnalysisOct 25, 2023192860010, 192862500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection