Ammonium metavanadate, 99.996%, (trace metal basis)
Ammonium metavanadate, 99.996%, (trace metal basis)
Ammonium metavanadate, 99.996%, (trace metal basis)
Ammonium metavanadate, 99.996%, (trace metal basis)
Thermo Scientific Chemicals

Ammonium metavanadate, 99.996%, (trace metal basis)

CAS: 7803-55-6 | H4NO3V | 116.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 194400500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7803-55-6
IUPAC Namevanadiumoylol amine
Molecular FormulaH4NO3V
InChI KeyYBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILESN.O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Appearance (Form)Powder or crystalline powder
Vanadium (V)42.4 to 44.6 %
Total trace metal impurities=<40 ppm
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0465054Certificate of AnalysisJul 31, 2024194400500, 194400100
A0458472Certificate of AnalysisDec 07, 2023194400500, 194400100
A0456891Certificate of AnalysisNov 03, 2023194400500, 194400100
A0452831Certificate of AnalysisMay 05, 2023194400500, 194400100
A0440003Certificate of AnalysisMar 23, 2022194400500, 194400100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection