Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3
Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3
Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3
Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3
Thermo Scientific Chemicals

Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 196151000
Price (EUR)
16,66
Online Exclusive
19,60
Save 2,94 (15%)
Each
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
16,66
Online Exclusive
19,60
Save 2,94 (15%)
Each
Add to cart
Specifications
CAS Max %2.1
CAS Min %1.9
CAS Max %97.1
CAS Min %96.9
CAS7440-02-0
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471143Certificate of AnalysisMay 17, 2025196155000, 196151000
A0468306Certificate of AnalysisFeb 04, 2025196155000, 196151000
A0463882Certificate of AnalysisJun 20, 2024196155000, 196151000
A0460480Certificate of AnalysisFeb 26, 2024196155000, 196151000
A0454440Certificate of AnalysisAug 25, 2023196155000, 196151000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Substances/mixtures corrosive to metal
Category 1
Skin Corrosion/Irritation
Category 1
Serious Eye Damage/Eye Irritation
Category 1
Carcinogenicity
Category 1A
Reproductive Toxicity
Category 1B
Chronic aquatic toxicity
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H314 - Causes severe skin burns and eye damage

H350i - May cause cancer by inhalation

H360D - May damage the unborn child

H412 - Harmful to aquatic life with long lasting effects

Physical hazards

H290 - May be corrosive to metals

Containing sensitiser @ < 1%

EUH208 - Contains Nickel. May produce an allergic reaction

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Additional EU labelling

Restricted to professional users

Share catalog number, name or link

1x1 image pixel for data collection