Thermo Scientific Chemicals

Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 198405000
500 g, Each
Thermo Scientific Chemicals

Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 198405000
500 g, Each
Quantity
Catalog number: 198405000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10450-60-9
IUPAC Name
tetrahydroxyiodous acid
Molecular Formula
H5IO6
InChI Key
TWLXDPFBEPBAQB-UHFFFAOYSA-N
SMILES
O[I](O)(O)(O)(O)=O
Identification
Positive for periodate
Insoluble matter
=<0.01 % (in water)
Sulfate (SO4)
=<200 ppm
Appearance (Color)
White to light yellow
Titration Iodometric
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text