Thermo Scientific™

Pierce™ Dimethylsulfoxide (DMSO), Sequencing Grade

Catalog number: 20688
Have Questions?
Thermo Scientific™

Pierce™ Dimethylsulfoxide (DMSO), Sequencing Grade

Catalog number: 20688
Have Questions?
Catalog Number
20688
Unit Size
950 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
20688950 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Dimethylsulfoxide (DMSO) is a sequencing-grade preparation with high purity for use in traditional amino acid analyzers and other high-volume applications.

This sequencing-grade dimethylsulfoxide is >99.5% pure DMSO. The economically priced 950 mL bottles are an excellent choice for protein and peptide applications that requiring large volumes of the water-miscible solvent. (For small-volume needs, consider the related 50 mL bottles of DMSO, Cat. No. 85190.) The purity test ensures that preparation is essentially devoid of amines. The formulation was originally developed for peptide sequencing applications with amino acid analyzers, which required high purity solvent and an affordable price. This DMSO is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.

Features of DMSO:

• High-purity (low amine) solvent for various protein, peptide and amino acid applications
• Packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps
• Suitable for HPLC detection, although it is not specifically tested for low UV-absorptivity

General properties of DMSO
• Alternative names: Methylsulfinylmethane, methyl sulfoxide
• Molecular formula: C2H6SO
• Molecular weight: 78.13
• Density: 1.101 g/mL
• CAS number: 67-68-5

Specifications of Thermo Scientific Pierce DMSO
• Particulates: Free from particulate matter
• Purity: > 99.5%
• Water content: < 0.2%

This DMSO is specially purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and to prolong the life of your equipment.

Related Products
Pierce™ Dimethylsulfoxide (DMSO), LC-MS Grade
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Item Name
Dimethylsulfoxide (DMSO)
Quantity
950 mL
Form
Liquid
Reagent Type
Solvent (Pure)
Product Line
Pierce™
For Use With
Sequencing, Liquid Chromatography

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text