Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | ART 300, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 300 μL |
Volume (Metric) Dispensing | 300μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 300 |
Length (English) | 2.3 in. |
Length (Metric) | 5.84 cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Micro Point |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Material | Polypropylene |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Pack of 960 |
Description | ART 200, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 200 μL |
Volume (Metric) Dispensing | 200μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 200 |
Length (English) | 2.32 in |
Length (Metric) | 5.88cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Universal |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Material | Polypropylene |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Pack of 960 |
Description | ART 100E, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 100 μL |
Volume (Metric) Dispensing | 100μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 100E |
Length (English) | 2 in. |
Length (Metric) | 5.07cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Micro Point |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Material | Polypropylene |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Pack of 960 |
Description | ART 300, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 300 μL |
Volume (Metric) Dispensing | 300μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 300 |
Length (English) | 2.3 in. |
Length (Metric) | 5.84 cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Micro Point |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Material | Polypropylene |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Case of 4800 |
Description | ART 100E, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 100 μL |
Volume (Metric) Dispensing | 100μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 100E |
Length (English) | 2 in. |
Length (Metric) | 5.07cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Micro Point |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Material | Polypropylene |
Fit for Purpose | Certified Quality |
Unit Size | Case of 4800 |
Description | ART 200, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 200 μL |
Volume (Metric) Dispensing | 200μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 200 |
Length (English) | 2.32 in |
Length (Metric) | 5.88cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Universal |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Material | Polypropylene |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Case of 4800 |
Description | ART 100, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 100 μL |
Volume (Metric) Dispensing | 100μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 100 |
Length (English) | 2.32 in |
Length (Metric) | 5.88cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Micro Point |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Material | Polypropylene |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Case of 4800 |
Description | ART 100, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 100 μL |
Volume (Metric) Dispensing | 100μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 100 |
Length (English) | 2.32 in |
Length (Metric) | 5.88cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Micro Point |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Material | Polypropylene |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Pack of 960 |
Description | ART 1000E, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 1000 μL |
Volume (Metric) Dispensing | 1000μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 1000E |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Universal |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Material | Polypropylene |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Case of 3200 |
Description | ART 1000E, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 1000 μL |
Volume (Metric) Dispensing | 1000μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 1000E |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Universal |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Material | Polypropylene |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Pack of 800 |
Catalog Number | Specifications | Unit Size | Description | Volume (Metric) | Tip Style | Fit for Purpose | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2070PK also known as 2070 | Pack of 960 | ART 300, Filtered, Sterile, Lift-off Lid Rack | 300 μL | Micro Point | Sterile & PCR Ready | 90.80 Online price 196.90 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2069PK also known as 2069 | Pack of 960 | ART 200, Filtered, Sterile, Lift-off Lid Rack | 200 μL | Universal | Sterile & PCR Ready | 211.30 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2065EPK also known as 2065E | Pack of 960 | ART 100E, Filtered, Sterile, Lift-off Lid Rack | 100 μL | Micro Point | Sterile & PCR Ready | 197.90 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2070 | Case of 4800 | ART 300, Filtered, Sterile, Lift-off Lid Rack | 300 μL | Micro Point | Sterile & PCR Ready | 454.00 Online price 863.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2065E | Case of 4800 | ART 100E, Filtered, Sterile, Lift-off Lid Rack | 100 μL | Micro Point | Certified Quality | 607.60 Online price 868.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2069 | Case of 4800 | ART 200, Filtered, Sterile, Lift-off Lid Rack | 200 μL | Universal | Sterile & PCR Ready | 649.60 Online price 928.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2065 | Case of 4800 | ART 100, Filtered, Sterile, Lift-off Lid Rack | 100 μL | Micro Point | Sterile & PCR Ready | 607.60 Online price 868.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2065PK also known as 2065 | Pack of 960 | ART 100, Filtered, Sterile, Lift-off Lid Rack | 100 μL | Micro Point | Sterile & PCR Ready | 197.90 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2079E | Case of 3200 | ART 1000E, Filtered, Sterile, Lift-off Lid Rack | 1000 μL | Universal | Sterile & PCR Ready | 480.20 Online price 686.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2079EPK also known as 2079E | Pack of 800 | ART 1000E, Filtered, Sterile, Lift-off Lid Rack | 1000 μL | Universal | Sterile & PCR Ready | 195.90 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features:
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes