Calcium chloride dihydrate, 99+%, for analysis
Calcium chloride dihydrate, 99+%, for analysis
Calcium chloride dihydrate, 99+%, for analysis
Calcium chloride dihydrate, 99+%, for analysis
Thermo Scientific Chemicals

Calcium chloride dihydrate, 99+%, for analysis

CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2077800101 kgPlastic Bottle
Catalog number 207780010
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS10035-04-8
IUPAC Namecalcium dihydrate dichloride
Molecular FormulaCaCl2H4O2
InChI KeyLLSDKQJKOVVTOJ-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Magnesium (Mg)=<50 ppm
Titration Complexometric>=99.0 %
Iron (Fe)=<5 ppm
Appearance (Form)Crystalline powder or crystals
Heavy metals (as Pb)=<5 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470511Certificate of AnalysisMay 17, 2025207780100, 207780010, 207780025
A0462140Certificate of AnalysisJun 05, 2024207780100, 207780010, 207780025
A0457953Certificate of AnalysisNov 21, 2023207780100, 207780010, 207780025
A0457020Certificate of AnalysisNov 08, 2023207780100, 207780010, 207780025
A0452886Certificate of AnalysisMay 10, 2023207780100, 207780010, 207780025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection