1x1 image pixel for data collection
Sodium chloride, 99.5%, for analysis
Sodium chloride, 99.5%, for analysis
Sodium chloride, 99.5%, for analysis
Sodium chloride, 99.5%, for analysis
Thermo Scientific Chemicals

Sodium chloride, 99.5%, for analysis

Sodium chloride, 99.5%, ClNa, CAS Number-7647-14-5, sodium chloride nacl, salt, common salt, rock salt, halite, purex, table salt, sodium chloride, saline, dendritis, 1kg, 99.4% min. (Argentometry), 10ppm max., 1461.0 deg.C, 50ppm max., 20ppm max., CHEBI:26710, White, 231-598-3, 13,280, 58.44 | CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic Bottle
Catalog number 207790010
Price (USD)
59.65
Special offer
Online exclusive
Ends: 26-Sep-2025
73.90
Save 14.25 (19%)
Each
In stock
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
59.65
Special offer
Online exclusive
Ends: 26-Sep-2025
73.90
Save 14.25 (19%)
Each
Add to cart
Chemical Identifiers
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<5 ppm
Total nitrogen (as N)=<0.001 %
Barium (Ba)=<10 ppm
Potassium (K)=<100 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2527966Certificate of AnalysisMay 24, 2025207790050, 207790250, 207790010
A0468368Certificate of AnalysisJan 31, 2025207790050, 207790250, 207790010
2404508Certificate of AnalysisJan 31, 2025207790050, 207790250, 207790010
2497961Certificate of AnalysisJan 31, 2025207790050, 207790250, 207790010
2407894Certificate of AnalysisJan 30, 2025207790050, 207790250, 207790010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection