Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company
Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company
Thermo Scientific Chemicals

Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company

CAS: 81133-20-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 208085000
Price (USD)
530.65
Special offer
Online exclusive
Ends: 26-Sep-2025
655.00
Save 124.35 (19%)
Each
In stock
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
530.65
Special offer
Online exclusive
Ends: 26-Sep-2025
655.00
Save 124.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS81133-20-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granules
Particle size8 to 20 mesh
Appearance (Color)Brown
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Gas Adsorbent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0464187Certificate of AnalysisJul 02, 2024208080025, 208085000, 208081000
A0451053Certificate of AnalysisMay 02, 2023208080025, 208085000, 208081000
A0451054Certificate of AnalysisMay 02, 2023208080025, 208085000, 208081000
A0448663Certificate of AnalysisApr 10, 2023208080025, 208085000, 208081000
A0414489Certificate of AnalysisDec 13, 2022208080025, 208085000, 208081000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection