Cobalt(II) nitrate hexahydrate, 99%, pure
Cobalt(II) nitrate hexahydrate, 99%, pure
Cobalt(II) nitrate hexahydrate, 99%, pure
Cobalt(II) nitrate hexahydrate, 99%, pure
Thermo Scientific Chemicals

Cobalt(II) nitrate hexahydrate, 99%, pure

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 213095000
Price (USD)
201.65
Special offer
Online exclusive
Ends: 26-Sep-2025
249.00
Save 47.35 (19%)
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
201.65
Special offer
Online exclusive
Ends: 26-Sep-2025
249.00
Save 47.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS10026-22-9
IUPAC Nameλ²-cobalt(2+) hexahydrate dinitrate
Molecular FormulaCoH12N2O12
InChI KeyQGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder or crystals or chips
Titration Complexometric>=98.5 %
Appearance (Color)Pink to red to brown
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469155Certificate of AnalysisJan 31, 2025213095000, 213090010, 213091000, 213090050
A0444035Certificate of AnalysisJan 31, 2025213095000, 213090010, 213091000, 213090050
A0463812Certificate of AnalysisJun 18, 2024213095000, 213090010, 213091000, 213090050
A0458020Certificate of AnalysisNov 30, 2023213095000, 213090010, 213091000, 213090050
A0426085Certificate of AnalysisNov 13, 2023213095000, 213090010, 213091000, 213090050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection