Ham's F-12 Nutrient Mix, powder
Save up to 29% on your order of cell culture products. View promotion details >
Product Image
Gibco™

Ham's F-12 Nutrient Mix, powder

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has sinceRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2170007510 x 1 L
2170001850 L
Catalog number 21700075
Price (USD)
58.65
Online Exclusive
62.00
Save 3.35 (5%)
Each
-
Add to cart
Quantity:
10 x 1 L
Customize this product
Price (USD)
58.65
Online Exclusive
62.00
Save 3.35 (5%)
Each
Add to cart
Ask our AI about this Product

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of Gibco™ F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This Ham's F-12 is modified as follows:
WithWithout
• L-glutamine• Sodium Bicarbonate
• Phenol Red 


The complete formulation is available.

Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-12 has led to a variety of improved formulations.

Gibco™ F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.
Specifications
Cell TypePrimary Rat Astrocytes
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco™
Product TypeHam's F-12 Nutrient Mixture
Quantity10 x 1 L
Shelf Life36 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormPowder
SterilitySterile-filtered
With AdditivesGlutamine, Phenol Red, Sodium Pyruvate
Without AdditivesNo HEPES, No Sodium Bicarbonate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Media Formulations
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3164236Certificate of AnalysisMay 02, 202521700075, 21700026, 21700018
3124521Certificate of AnalysisApr 05, 202521700075, 21700026, 21700018
3129360Certificate of AnalysisFeb 19, 202521700075, 21700026, 21700018
2792063Certificate of AnalysisJan 31, 202521700075, 21700026, 21700018
2482972Certificate of AnalysisJan 29, 202521700075, 21700018
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

Expiration date of most Gibco reconstituted dry format media (AGT or DPM) has not been established; end users should assess performance and stability of this reconstituted media in a system that is relevant to their process. A dry format product (either DPM or AGT) will age upon storage and while we may not be able to detect which component(s) degrade since we can't measure every component, that doesn't mean that the reconstituted liquid will have the same stability as a fresh liquid made by direct weigh methods.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (4)

Citations & References
Abstract
Astrocytes in culture produce anandamide and other acylethanolamides.
Authors: Walter Lisa; Franklin Allyn; Witting Anke; Moller Thomas; Stella Nephi;
Journal:J Biol Chem
PubMed ID:11916961
'Anandamide (arachidonylethanolamide) is an endocannabinoid that belongs to the acylethanolamide lipid family. It is produced by neurons in a calcium-dependent manner and acts through cannabinoid CB1 receptors. Other members of the acylethanolamide lipid family are also produced by neurons and act through G-protein-coupled receptors: homo-gamma-linolenylethanolamide (HEA) and docosatetraenylethanolamide (DEA) act ... More
The Slp homology domain of synaptotagmin-like proteins 1-4 and Slac2 functions as a novel Rab27A binding domain.
Authors: Kuroda Taruho S; Fukuda Mitsunori; Ariga Hiroyoshi; Mikoshiba Katsuhiko;
Journal:J Biol Chem
PubMed ID:11773082
rab27A, which encodes a small GTP-binding protein, was recently identified as a gene in which mutations caused human hemophagocytic syndrome (Griscelli syndrome) and ashen mice, which exhibit defects in melanosome transport as well as in regulated granule exocytosis in cytotoxic T lymphocytes. However, little is known about the molecular mechanism ... More
High density lipoprotein binding to scavenger receptor, Class B, type I activates endothelial nitric-oxide synthase in a ceramide-dependent manner.
Authors: Li Xiang-An; Titlow William B; Jackson Brian A; Giltiay Nathalia; Nikolova-Karakashian Mariana; Uittenbogaard Annette; Smart Eric J;
Journal:J Biol Chem
PubMed ID:11792700
Recently it has been demonstrated that high density lipoprotein (HDL) binding to scavenger receptors, class B, type I (SR-BI) stimulates endothelial nitric-oxide synthase (eNOS) activity. In the present studies we used a Chinese hamster ovary cell system and a human microvascular endothelial cell line to confirm that HDL stimulates eNOS ... More
Neuropeptide AF and FF modulation of adipocyte metabolism. Primary insights from functional genomics and effects on beta-adrenergic responsiveness.
Authors:Lefrere I, De Coppet P, Camelin JC, Le Lay S, Mercier N, Elshourbagy N, Bril A, Berrebi-Bertrand I, Feve B, Krief S
Journal:J Biol Chem
PubMed ID:12149260
The presence of a neuropeptide AF and FF receptor (NPFF-R2) mRNA in human adipose tissue (Elshourbagy, N. A., Ames, R. S., Fitzgerald, L. R., Foley, J. J., Chambers, J. K., Szekeres, P. G., Evans, N. A., Schmidt, D. B., Buckley, P. T., Dytko, G. M., Murdock, P. R., Tan, K. ... More
4 total citations

Andere Produkte



Share catalog number, name or link