Thermo Scientific™
Temperature Chambers for HAAKE™ MARS™ Rheometers
Catalog number: 222-2176
Have Questions?
Thermo Scientific™
Temperature Chambers for HAAKE™ MARS™ Rheometers
Catalog number: 222-2176
Have Questions?
The Thermo Scientific™ HAAKE™ MARS™ rheometer platforms feature temperature chambers, which ensure reliable and accurate sample temperature during rheological measurement of melts and solids over a wide temperature range. 
Temperature chambers for HAAKE MARS 40/60 and HAAKE MARS iQ Air Rheometers are available.
Both solutions offer advantages such as:
  • Patented combination of convection and radiation heat transfer for fast temperature changes and homogeneous temperature distribution 
  • Fast sample loading, trimming and cleaning due to easy sample access
  • Wide  portfolio of measuring geometries
Catalog Number
222-2176
Unit Size
Each
Description
Sentmanat Extensional Rheometer (SER) tool from Xpansion Instruments for extensional rheological measurements of polymer films
Price (USD)
Full specifications
TypeSentmanat Extensional Rheomete (SER) Tool
DescriptionSentmanat Extensional Rheometer (SER) tool from Xpansion Instruments for extensional rheological measurements of polymer films
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Controlled Test Chamber (CTC) for HAAKE MARS Rheometer (models I, III, 40, 60)
Price (USD)
Full specifications
TypeControlled Test Chamber
DescriptionControlled Test Chamber (CTC) for HAAKE MARS Rheometer (models I, III, 40, 60)
Temperature Range (Metric)30°C to 600°C or Low-Temperature Option: down to –150°C with liquid nitrogen device
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Self-centering and self-adjusting clamps for Dynamic Mechanical Thermal Analysis (DMTA) of solid specimen
Price (USD)
Full specifications
TypeClamps
DescriptionSelf-centering and self-adjusting clamps for Dynamic Mechanical Thermal Analysis (DMTA) of solid specimen
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Optional “low temperature”, premium version with low energy consumption, down to -150 °C
Price (USD)
Full specifications
TypeLow Temperature Option for CTC
DescriptionOptional “low temperature”, premium version with low energy consumption, down to -150 °C
Unit SizeEach
Catalog Number
Unit Size
Each
Description
UV measuring cell for CTC HAAKE MARS
Price (USD)
Full specifications
TypeUV Measuring Cell
DescriptionUV measuring cell for CTC HAAKE MARS
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Temperature Chamber for HAAKE MARS iQ Air Rheometers
Price (USD)
Full specifications
TypeControlled Test Chamber
DescriptionTemperature Chamber for HAAKE MARS iQ Air Rheometers
Temperature Range (Metric) 30 to 600°C or Low-Temperature Option: down to -150°C with liquid nitrogen device
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
222-2176Full specifications
EachSentmanat Extensional Rheometer (SER) tool from Xpansion Instruments for extensional rheological measurements of polymer filmsRequest A Quote
TypeSentmanat Extensional Rheomete (SER) Tool
DescriptionSentmanat Extensional Rheometer (SER) tool from Xpansion Instruments for extensional rheological measurements of polymer films
Unit SizeEach
222-1913Full specifications
EachControlled Test Chamber (CTC) for HAAKE MARS Rheometer (models I, III, 40, 60)Request A Quote
TypeControlled Test Chamber
DescriptionControlled Test Chamber (CTC) for HAAKE MARS Rheometer (models I, III, 40, 60)
Temperature Range (Metric)30°C to 600°C or Low-Temperature Option: down to –150°C with liquid nitrogen device
Unit SizeEach
222-2231Full specifications
EachSelf-centering and self-adjusting clamps for Dynamic Mechanical Thermal Analysis (DMTA) of solid specimenRequest A Quote
TypeClamps
DescriptionSelf-centering and self-adjusting clamps for Dynamic Mechanical Thermal Analysis (DMTA) of solid specimen
Unit SizeEach
222-1730Full specifications
EachOptional “low temperature”, premium version with low energy consumption, down to -150 °CRequest A Quote
TypeLow Temperature Option for CTC
DescriptionOptional “low temperature”, premium version with low energy consumption, down to -150 °C
Unit SizeEach
603-0315Full specifications
EachUV measuring cell for CTC HAAKE MARSRequest A Quote
TypeUV Measuring Cell
DescriptionUV measuring cell for CTC HAAKE MARS
Unit SizeEach
222-2468Full specifications
EachTemperature Chamber for HAAKE MARS iQ Air RheometersRequest A Quote
TypeControlled Test Chamber
DescriptionTemperature Chamber for HAAKE MARS iQ Air Rheometers
Temperature Range (Metric) 30 to 600°C or Low-Temperature Option: down to -150°C with liquid nitrogen device
Unit SizeEach
Showing 6 of 6
Controlled test chamber (CTC) for HAAKE MARS 40/60 Rheometer
Two independent halves provide numerous advantages for convenient sample handling and optimal access:
  • Each half can be moved independently backward and sideways – no special tools needed
  • When closed, the two halves automatically center themselves around the sample
  • Each chamber is equipped with a window to observe the sample during measurement
  • A “park” position feature allows the attachment of another temperature control unit without removing the CTC
  • In the “park” position, the temperature inside the CTC can be maintained while the geometry is cleaned or the next sample is loaded
No time is wasted waiting for the CTC to reach its set temperature
  • The CTC's temperature range spans from 30°C to 600°C and can be expanded down to -150°C with an optional liquid nitrogen device
Temperature chamber for HAAKE MARS iQ Rheometers
Oven with compact design
  • Easily installed without additional alignment when changing between different temperature modules
  • Two halves of the chamber open in a wide angle for excellent access to the measuring geometries
  • Optional camera with illumination for taking pictures or videos during sample measurement
The temperature range spans from 30°C to 450°C, expandable down to:
  • -150°C with an optional liquid nitrogen device. 
  • -50°C with a cold gas chiller
A portfolio of varies measuring geometries is available depending on the oven solution: 
  • Comprehensive range of parallel plate,  – cone and plate or coaxial cylinder measuring geometries
  • Various  clamp setups for Dynamic Mechanical Thermal Analysis (DMTA) 
  • Sentmanat Extension Rheometer (SER) tool from Xpansion Instruments for extensional rheological investigations on polymer films
  • Various tribology setups, e.g. ball on three balls for determining friction and wear of lubricants over a wide temperature range  