Search Thermo Fisher Scientific
Type | Clamps |
Description | Self-centering and self-adjusting clamps for Dynamic Mechanical Thermal Analysis (DMTA) of solid specimen |
Unit Size | Each |
Type | Controlled Test Chamber |
Description | Controlled Test Chamber (CTC) for HAAKE MARS Rheometer (models I, III, 40, 60) |
Temperature Range (Metric) | 30°C to 600°C or Low-Temperature Option: down to –150°C with liquid nitrogen device |
Unit Size | Each |
Type | Low Temperature Option for CTC |
Description | Optional “low temperature”, premium version with low energy consumption, down to -150 °C |
Unit Size | Each |
Type | Sentmanat Extensional Rheomete (SER) Tool |
Description | Sentmanat Extensional Rheometer (SER) tool from Xpansion Instruments for extensional rheological measurements of polymer films |
Unit Size | Each |
Type | UV Measuring Cell |
Description | UV measuring cell for CTC HAAKE MARS |
Unit Size | Each |
Type | Controlled Test Chamber |
Description | Temperature Chamber for HAAKE MARS iQ Air Rheometers |
Temperature Range (Metric) | 30 to 600°C or Low-Temperature Option: down to -150°C with liquid nitrogen device |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
222-2231 | Each | Self-centering and self-adjusting clamps for Dynamic Mechanical Thermal Analysis (DMTA) of solid specimen | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
222-1913 | Each | Controlled Test Chamber (CTC) for HAAKE MARS Rheometer (models I, III, 40, 60) | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
222-1730 | Each | Optional “low temperature”, premium version with low energy consumption, down to -150 °C | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
222-2176 | Each | Sentmanat Extensional Rheometer (SER) tool from Xpansion Instruments for extensional rheological measurements of polymer films | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
603-0315 | Each | UV measuring cell for CTC HAAKE MARS | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
222-2468 | Each | Temperature Chamber for HAAKE MARS iQ Air Rheometers | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|