Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store and transport solids, powders and liquids using Thermo Scientific™ Nalgene™ Wide-Mouth Autoclavable Polypropylene Carboys with Handles. These durable natural polypropylene (PP) carboys feature a convenient wide-mouth opening to promote ease of cleaning and dispensing. The wide-shoulder handles enable easy carrying and pouring, even with gloved hands.
Autoclavable | Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Material | Polypropylene |
Closure Size | 100-415 mm |
Color | White |
Description | Autoclavable Wide-Mouth Carboy with Handles |
Diameter (Metric) Outer | 282 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Graduations | Graduated to contain in 5 L/1 gal. increments |
Height (Metric) With Closure | 483 mm |
Includes | Handles |
Material | Polypropylene |
Mouth Style | Wide |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Handle |
Sterility | Non-sterile |
Volume (English) Brim | 5.5 gal. |
Volume (Metric) Brim | 22.5 L |
Unit Size | Each |
Autoclavable | Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Closure Material | Polypropylene |
Closure Size | 100-415 mm |
Color | White |
Description | Autoclavable Wide-Mouth Carboy with Handles |
Diameter (Metric) Outer | 250 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Graduations | Graduated to contain in 5 L/1 gal. increments |
Height (Metric) With Closure | 343 mm |
Includes | Handles |
Material | Polypropylene |
Mouth Style | Wide |
No. per Case | 6/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Handle |
Sterility | Non-sterile |
Volume (English) Brim | 3 gal. |
Volume (Metric) Brim | 12 L |
Unit Size | Case of 6 |
Autoclavable | Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Closure Material | Polypropylene |
Closure Size | 100-415 mm |
Color | White |
Description | Autoclavable Wide-Mouth Carboy with Handles |
Diameter (Metric) Outer | 250 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Graduations | Graduated to contain in 5 L/1 gal. increments |
Height (Metric) With Closure | 343 mm |
Includes | Handles |
Material | Polypropylene |
Mouth Style | Wide |
No. per Case | 6/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Handle |
Sterility | Non-sterile |
Volume (English) Brim | 3 gal. |
Volume (Metric) Brim | 12 L |
Unit Size | Each |
Autoclavable | Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Material | Polypropylene |
Closure Size | 100-415 mm |
Color | White |
Description | Autoclavable Wide-Mouth Carboy with Handles |
Diameter (Metric) Outer | 282 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Graduations | Graduated to contain in 5 L/1 gal. increments |
Height (Metric) With Closure | 483 mm |
Includes | Handles |
Material | Polypropylene |
Mouth Style | Wide |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Handle |
Sterility | Non-sterile |
Volume (English) Brim | 5.5 gal. |
Volume (Metric) Brim | 22.5 L |
Unit Size | Case of 4 |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2235-0050PK | Each | 20 L | 240.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2235-0020 | Case of 6 | 10 L | 924.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2235-0020PK | Each | 10 L | 174.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2235-0050 | Case of 4 | 20 L | 854.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.