Rosolic acid
Rosolic acid
Rosolic acid
Thermo Scientific Chemicals

Rosolic acid

CAS: 603-45-2 | C19H14O3 | 290.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 229830250
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
In stock
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS603-45-2
IUPAC Name4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
Molecular FormulaC19H14O3
InChI KeyFYEHYMARPSSOBO-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
UV Lambda max480 to 486 nm (in ethanol + 2ml 1N HCl)
Appearance (Form)Crystalline powder
Visual transition intervalfrom pH 6.2 (yellow) to pH 8.2 (red-pink)
Appearance (Color)Red-brown to brown
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468813Certificate of AnalysisJan 31, 2025229830250, 229831000, 229835000
A0467928Certificate of AnalysisJan 31, 2025229830250, 229831000, 229835000
A0465510Certificate of AnalysisAug 30, 2024229830250, 229831000, 229835000
A0462557Certificate of AnalysisMay 16, 2024229830250, 229831000, 229835000
A0392727Certificate of AnalysisDec 15, 2022229830250, 229831000, 229835000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection