Thermo Scientific™

FITC-Casein for Pierce™ Fluorescent Protease Assay Kit

Catalog number: 23267
Have Questions?
Thermo Scientific™

FITC-Casein for Pierce™ Fluorescent Protease Assay Kit

Catalog number: 23267
Have Questions?
Catalog Number
23267
Unit Size
2.5 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
232672.5 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
FTC-Casein, 2.5 mg, lyophilized, FRET/FP Certified Grade, for use with the Pierce Fluorescent Quantitative Protease Assay Kit.

Related Products
Pierce™ Colorimetric Protease Assay Kit
Pierce™ Fluorescent Protease Assay Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay
Substrate
Detection Method
Fluorescence
Immunoassay Kit Format
96-well Plate
Label Type
Classic Dye
Product Line
Pierce™
Product Type
Protein Activity Assay
Substrate
FITC-Casein
Substrate Properties
Protein-Based Substrate
Substrate Type
Protease Substrate
For Use With (Application)
Fluorescence Polarization, FRET
For Use With (Equipment)
Microplate Reader
Quantity
2.5 mg
Target
Proteases

Contents & Storage

Upon receipt store FTC-Casein at 4°C. Product shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text