Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.
Product Type | Tubing Set |
Autoclavable | Autoclavable 50 times at 121°C, 1 bar, 20 min |
Description | Tubing set for standard dispensing cassette (tubing length 40cm) |
Material | Polypropylene Tip, Silicone Tubing |
Volume (Metric) | 5 to 2500 μL |
For Use With (Equipment) | Multidrop Combi, Multidrop 384, Multidrop DW |
Volume (Metric) Dead | <7 mL |
Diameter (Metric) | Tip: 0.5 mm; Tubing: 1.3 mm |
Unit Size | Each |
Product Type | Long Tubing Set |
Autoclavable | Autoclavable 50 times at 121°C, 1 bar, 20 min |
Description | Long tubing set for standard dispensing cassette (tubing length 200 cm) |
Material | Polypropylene Tip, Silicone Tubing |
Volume (Metric) | 5 to 2500 μL |
For Use With (Equipment) | Multidrop Combi, Multidrop 384, Multidrop DW |
Volume (Metric) Dead | <24 mL |
Diameter (Metric) | Tip: 0.5 mm; Tubing: 1.3 mm |
Unit Size | Each |
Product Type | Tube Dispensing Cassette |
Autoclavable | Autoclavable 50 times at 121°C, 1 bar, 20 min |
Description | Standard tube dispensing cassette |
Material | Polypropylene Tip, Silicone Tubing |
Volume (Metric) | 5 to 2500 μL |
For Use With (Equipment) | Multidrop Combi, Multidrop 384, Multidrop DW |
Volume (Metric) Dead | <7 mL |
Diameter (Metric) | Tip: 0.5 mm; Tubing: 1.3 mm |
Unit Size | Each |
Product Type | Tube Dispensing Cassette |
Autoclavable | Autoclavable 50 times at 121°C, 1 bar, 20 min |
Description | Standard tube dispensing cassette 5-pack |
Material | Polypropylene Tip, Silicone Tubing |
Volume (Metric) | 5 to 2500 μL |
For Use With (Equipment) | Multidrop Combi, Multidrop 384, Multidrop DW |
Volume (Metric) Dead | <7 mL |
Diameter (Metric) | Tip: 0.5 mm; Tubing: 1.3 mm |
Unit Size | Each |
Product Type | Tube Dispensing Cassette |
Autoclavable | Autoclavable 50 times at 121°C, 1 bar, 20 min |
Description | Standard tube dispensing cassette 10-pack |
Material | Polypropylene Tip, Silicone Tubing |
Volume (Metric) | 5 to 2500 μL |
For Use With (Equipment) | Multidrop Combi, Multidrop 384, Multidrop DW |
Volume (Metric) Dead | <7 mL |
Diameter (Metric) | Tip: 0.5 mm; Tubing: 1.3 mm |
Unit Size | Each |
Product Type | Tube Dispensing Cassette |
Autoclavable | Autoclavable 50 times at 121°C, 1 bar, 20 min |
Description | Long standard tube dispensing cassette (tubing length 200cm) |
Material | Polypropylene Tip, Silicone Tubing |
Volume (Metric) | 5 to 2500 μL |
For Use With (Equipment) | Multidrop Combi, Multidrop 384, Multidrop DW |
Volume (Metric) Dead | <7 mL |
Diameter (Metric) | Tip: 0.5 mm; Tubing: 1.3 mm |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
24070290 | Each | Tubing set for standard dispensing cassette (tubing length 40cm) | 433.80 Online price 482.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
24070297 | Each | Long tubing set for standard dispensing cassette (tubing length 200 cm) | 572.00 Online price 714.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
24072670 | Each | Standard tube dispensing cassette | 887.40 Online price 986.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
24072671 | Each | Standard tube dispensing cassette 5-pack | 3,628.80 Online price 4,032.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
24072672 | Each | Standard tube dispensing cassette 10-pack | 5,648.40 Online price 6,276.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
24072677 | Each | Long standard tube dispensing cassette (tubing length 200cm) | 1,300.00 Online price 1,444.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information:
Cat Nos 24070297 and 24070290 include tip combs.