Thermo Scientific™

Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories

Catalog number: 24072677
Have Questions?
Thermo Scientific™

Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories

Catalog number: 24072677
Have Questions?

Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.

 
Catalog Number
24072677
Unit Size
Each
Description
Long standard tube dispensing cassette (tubing length 200cm)
Price (USD)
Availability
-
Full specifications
Quantity
Product TypeTube Dispensing Cassette
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionLong standard tube dispensing cassette (tubing length 200cm)
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<7 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Tubing set for standard dispensing cassette (tubing length 40cm)
Price (USD)
Availability
-
Full specifications
Quantity
Product TypeTubing Set
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionTubing set for standard dispensing cassette (tubing length 40cm)
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<7 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Long tubing set for standard dispensing cassette (tubing length 200 cm)
Price (USD)
Availability
-
Full specifications
Quantity
Product TypeLong Tubing Set
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionLong tubing set for standard dispensing cassette (tubing length 200 cm)
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<24 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Standard tube dispensing cassette
Price (USD)
Availability
-
Full specifications
Quantity
Product TypeTube Dispensing Cassette
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionStandard tube dispensing cassette
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<7 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Standard tube dispensing cassette 5-pack
Price (USD)
Availability
-
Full specifications
Quantity
Product TypeTube Dispensing Cassette
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionStandard tube dispensing cassette 5-pack
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<7 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Standard tube dispensing cassette 10-pack
Price (USD)
Availability
-
Full specifications
Quantity
Product TypeTube Dispensing Cassette
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionStandard tube dispensing cassette 10-pack
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<7 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
24072677Full specifications
EachLong standard tube dispensing cassette (tubing length 200cm)
-
Product TypeTube Dispensing Cassette
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionLong standard tube dispensing cassette (tubing length 200cm)
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<7 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
24070290Full specifications
EachTubing set for standard dispensing cassette (tubing length 40cm)
-
Product TypeTubing Set
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionTubing set for standard dispensing cassette (tubing length 40cm)
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<7 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
24070297Full specifications
EachLong tubing set for standard dispensing cassette (tubing length 200 cm)
-
Product TypeLong Tubing Set
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionLong tubing set for standard dispensing cassette (tubing length 200 cm)
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<24 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
24072670Full specifications
EachStandard tube dispensing cassette
-
Product TypeTube Dispensing Cassette
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionStandard tube dispensing cassette
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<7 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
24072671Full specifications
EachStandard tube dispensing cassette 5-pack
-
Product TypeTube Dispensing Cassette
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionStandard tube dispensing cassette 5-pack
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<7 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
24072672Full specifications
EachStandard tube dispensing cassette 10-pack
-
Product TypeTube Dispensing Cassette
AutoclavableAutoclavable 50 times at 121°C, 1 bar, 20 min
DescriptionStandard tube dispensing cassette 10-pack
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Volume (Metric) Dead<7 mL
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
Unit SizeEach
Showing 6 of 6

Ordering Information:

Cat Nos 24070297 and 24070290 include tip combs.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text