Informational:Inquire about OEM or Commercial Supply version of this product here.

KingFisher™ Flex Purification System

Catalog number: 24074411
Have Questions?

KingFisher™ Flex Purification System

Catalog number: 24074411
Have Questions?
Prepare samples efficiently from a variety of materials with Thermo Scientific KingFisher Flex Purification Systems, which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.
 
Catalog Number
24074411
Unit Size
Each
Description
KingFisher Flex 96 PCR head and heating block
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionKingFisher Flex 96 PCR head and heating block
Run Time20 min.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
KingFisher Flex 96 PCR Head
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionKingFisher Flex 96 PCR Head
Run Time20 min.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
KingFisher Flex 96 Deep Well head and deep well and KF heating blocks
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionKingFisher Flex 96 Deep Well head and deep well and KF heating blocks
Run Time20 min.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
KingFisher Flex 96 KF head
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionKingFisher Flex 96 KF head
Run Time20 min.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
KingFisher Flex 96 KF head and heating block
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionKingFisher Flex 96 KF head and heating block
Run Time20 min.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
KingFisher Flex 24 Deep Well head and heating block
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionKingFisher Flex 24 Deep Well head and heating block
Run Time20 min.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
KingFisher Flex 96 PCR heating block
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionKingFisher Flex 96 PCR heating block
Run Time20 min.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
KingFisher 96 Plate
Price (USD)
Availability
-
Full specifications
Quantity
-
Volume (Metric) Processing20 to 200 μL
Plates per Deck24 or 96
Format96-well Plate
Capacity96 Samples
DescriptionKingFisher 96 Plate
Collection Efficiency of Particles> 95%
DisplaySTART / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / four cursor keys / LCD
For Use With (Application)Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating TemperatureFrom +5°C above ambient temperature to +115°C
Magnetic Rods96 in one frame; Interchangeable magnet heads
Particle Size (Metric)ca. >1 μm
Plate Type24 or 96-well plates: KingFisher 24 Deep Well plate (200 to 5000 μL), Microtiter 96 Deep Well plate (50 to 1000 μL), KingFisher 96 plate (20 to 200 μL), PCR plate (20 to 100 μL)
Depth (English)23.6 in.
Depth (Metric)60 cm
Height (English)15 in.
Height (Metric)38 cm
Weight (English)62 lb.
Weight (Metric)28 kg
Width (English)26.8 in.
Width (Metric)68 cm
Unit SizeEach
Catalog Number
Unit Size
System + SmartStart + 1 yr ext warranty
Description
KingFisher 96 Plate, Extended Warranty Package
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric) Processing20 to 200 μL
Plates per Deck24 or 96
Format96-well Plate
Capacity96 Samples
DescriptionKingFisher 96 Plate, Extended Warranty Package
Collection Efficiency of Particles> 95%
DisplaySTART / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / Four cursor keys / LCD
For Use With (Application)Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating TemperatureFrom +5°C above ambient temperature to +115°C
Magnetic Rods96 in one frame; Interchangeable magnet heads
Particle Size (Metric)ca. >1 μm
Plate Type24 or 96-well plates; KingFisher 24 Deep Well plate (200 to 5000 μL); Microtiter 96 Deep Well plate (50 to 1000 μμL); KingFisher 96 plate (20 to 200 μL);PCR plate (20 to 100 μL)
Depth (English)23.6 in.
Depth (Metric)60 cm
Height (English)15 in.
Height (Metric)38 cm
Weight (English)62 lb.
Weight (Metric)28 kg
Width (English)26.8 in.
Width (Metric)68 cm
Customer Education1 day at customer site
Total Service Coverage1 year manufacturer warranty and 1 year of field service with PM
Unit SizeSystem + SmartStart + 1 yr ext warranty
Catalog Number
Unit Size
System + SmartStart
Description
KingFisher 96 Plate with SmartStart
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric) Processing20 to 200 μL
Plates per Deck24 or 96
Format96-well Plate
Capacity96 Samples
DescriptionKingFisher 96 Plate with SmartStart
Collection Efficiency of Particles> 95%
DisplaySTART / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / Four cursor keys / LCD
For Use With (Application)Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating TemperatureFrom +5°C above ambient temperature to +115°C
Magnetic Rods96 in one frame; Interchangeable magnet heads
Particle Size (Metric)ca. >1 μm
Plate Type24 or 96-well plates; KingFisher 24 Deep Well plate (200 to 5000 μL); Microtiter 96 Deep Well plate (50 to 1000 μL); KingFisher 96 plate (20 to 200 μL); PCR plate (20 to 100 μL)
Depth (English)23.6 in.
Depth (Metric)60 cm
Height (English)15 in.
Height (Metric)38 cm
Weight (English)62 lb.
Weight (Metric)28 kg
Width (English)26.8 in.
Width (Metric)68 cm
Customer Education1 day at customer site
Total Service Coverage1 year manufacturer warranty
Unit SizeSystem + SmartStart
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
24074411Full specifications
EachKingFisher Flex 96 PCR head and heating block
-
DescriptionKingFisher Flex 96 PCR head and heating block
Run Time20 min.
Unit SizeEach
24074410Full specifications
EachKingFisher Flex 96 PCR Head
-
DescriptionKingFisher Flex 96 PCR Head
Run Time20 min.
Unit SizeEach
24074431Full specifications
EachKingFisher Flex 96 Deep Well head and deep well and KF heating blocks
-
DescriptionKingFisher Flex 96 Deep Well head and deep well and KF heating blocks
Run Time20 min.
Unit SizeEach
24074420Full specifications
EachKingFisher Flex 96 KF head
-
DescriptionKingFisher Flex 96 KF head
Run Time20 min.
Unit SizeEach
24074421Full specifications
EachKingFisher Flex 96 KF head and heating block
-
DescriptionKingFisher Flex 96 KF head and heating block
Run Time20 min.
Unit SizeEach
24074441Full specifications
EachKingFisher Flex 24 Deep Well head and heating block
-
DescriptionKingFisher Flex 24 Deep Well head and heating block
Run Time20 min.
Unit SizeEach
24075410Full specifications
EachKingFisher Flex 96 PCR heating block
-
DescriptionKingFisher Flex 96 PCR heating block
Run Time20 min.
Unit SizeEach
5400620Full specifications
EachKingFisher 96 PlateRequest A Quote--
Volume (Metric) Processing20 to 200 μL
Plates per Deck24 or 96
Format96-well Plate
Capacity96 Samples
DescriptionKingFisher 96 Plate
Collection Efficiency of Particles> 95%
DisplaySTART / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / four cursor keys / LCD
For Use With (Application)Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating TemperatureFrom +5°C above ambient temperature to +115°C
Magnetic Rods96 in one frame; Interchangeable magnet heads
Particle Size (Metric)ca. >1 μm
Plate Type24 or 96-well plates: KingFisher 24 Deep Well plate (200 to 5000 μL), Microtiter 96 Deep Well plate (50 to 1000 μL), KingFisher 96 plate (20 to 200 μL), PCR plate (20 to 100 μL)
Depth (English)23.6 in.
Depth (Metric)60 cm
Height (English)15 in.
Height (Metric)38 cm
Weight (English)62 lb.
Weight (Metric)28 kg
Width (English)26.8 in.
Width (Metric)68 cm
Unit SizeEach
A39952Full specifications
System + SmartStart + 1 yr ext warrantyKingFisher 96 Plate, Extended Warranty Package
-
Volume (Metric) Processing20 to 200 μL
Plates per Deck24 or 96
Format96-well Plate
Capacity96 Samples
DescriptionKingFisher 96 Plate, Extended Warranty Package
Collection Efficiency of Particles> 95%
DisplaySTART / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / Four cursor keys / LCD
For Use With (Application)Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating TemperatureFrom +5°C above ambient temperature to +115°C
Magnetic Rods96 in one frame; Interchangeable magnet heads
Particle Size (Metric)ca. >1 μm
Plate Type24 or 96-well plates; KingFisher 24 Deep Well plate (200 to 5000 μL); Microtiter 96 Deep Well plate (50 to 1000 μμL); KingFisher 96 plate (20 to 200 μL);PCR plate (20 to 100 μL)
Depth (English)23.6 in.
Depth (Metric)60 cm
Height (English)15 in.
Height (Metric)38 cm
Weight (English)62 lb.
Weight (Metric)28 kg
Width (English)26.8 in.
Width (Metric)68 cm
Customer Education1 day at customer site
Total Service Coverage1 year manufacturer warranty and 1 year of field service with PM
Unit SizeSystem + SmartStart + 1 yr ext warranty
A39966Full specifications
System + SmartStartKingFisher 96 Plate with SmartStart
-
Volume (Metric) Processing20 to 200 μL
Plates per Deck24 or 96
Format96-well Plate
Capacity96 Samples
DescriptionKingFisher 96 Plate with SmartStart
Collection Efficiency of Particles> 95%
DisplaySTART / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / Four cursor keys / LCD
For Use With (Application)Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating TemperatureFrom +5°C above ambient temperature to +115°C
Magnetic Rods96 in one frame; Interchangeable magnet heads
Particle Size (Metric)ca. >1 μm
Plate Type24 or 96-well plates; KingFisher 24 Deep Well plate (200 to 5000 μL); Microtiter 96 Deep Well plate (50 to 1000 μL); KingFisher 96 plate (20 to 200 μL); PCR plate (20 to 100 μL)
Depth (English)23.6 in.
Depth (Metric)60 cm
Height (English)15 in.
Height (Metric)38 cm
Weight (English)62 lb.
Weight (Metric)28 kg
Width (English)26.8 in.
Width (Metric)68 cm
Customer Education1 day at customer site
Total Service Coverage1 year manufacturer warranty
Unit SizeSystem + SmartStart
Showing 10 of 10

Find a flexible solution for different types of sample processing needs. With 24 configurations, customers can raise the processing volume up to 5mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.

• Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
• High throughput system processes up to 96 samples in less than 20 minutes.
• Expands sample volume by 10-fold with the 24-well format.
• Open and flexible system lets the customer use anymagnetic particle-based kit tomeet the application demands.
• Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation andmodification.
• Ready-made protocols for different type of applications are available.

Recommended for:
• Genomics and proteomics
• Target identification
• Veterinary assays
• Biomarker discovery
• Quality control
• Forensics

For Laboratory Use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text