Diphosgene, 99%, Thermo Scientific Chemicals
Diphosgene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphosgene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
Catalog number 243310100
Price (USD)/ Each
68.65
Online exclusive
75.90 
Save 7.25 (10%)
-
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
68.65
Online exclusive
75.90 
Save 7.25 (10%)
Add to cart
Diphosgene, 99%, Thermo Scientific Chemicals
Catalog number243310100
Price (USD)/ Each
68.65
Online exclusive
75.90 
Save 7.25 (10%)
-
Add to cart
Chemical Identifiers
CAS503-38-8
IUPAC Nametrichloromethyl carbonochloridate
Molecular FormulaC2Cl4O2
InChI KeyHCUYBXPSSCRKRF-UHFFFAOYSA-N
SMILESClC(=O)OC(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4560 to 1.4590 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only