Water, pure, demineralized
Water, pure, demineralized
Water, pure, demineralized
Water, pure, demineralized
Thermo Scientific Chemicals

Water, pure, demineralized

CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
20 L
Packaging:
View all
Plastic Can
Catalog number 276010200
Price (USD)
266.65
Online Exclusive
296.00
Save 29.35 (10%)
Each
In stock
Add to cart
Quantity:
20 L
Packaging:
Plastic Can
Request bulk or custom format
Price (USD)
266.65
Online Exclusive
296.00
Save 29.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Conductivity=<5.0 µS/cm
pH6 to 8
Silicon dioxide (SiO2)=<1.0 ppm
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473341Certificate of AnalysisJun 12, 2025276010200, 276010050
A0473339Certificate of AnalysisJun 12, 2025276010200, 276010050
A0473340Certificate of AnalysisJun 12, 2025276010200, 276010050
A0467000Certificate of AnalysisNov 01, 2024276010200, 276010050
A0457204Certificate of AnalysisNov 20, 2023276010200, 276010050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection