3-Amino-3-phenyl-1-propanol, 94%
3-Amino-3-phenyl-1-propanol, 94%
3-Amino-3-phenyl-1-propanol, 94%
3-Amino-3-phenyl-1-propanol, 94%
Thermo Scientific Chemicals

3-Amino-3-phenyl-1-propanol, 94%

CAS: 14593-04-5 | C9H13NO | 151.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
292935000500 mgGlass bottle
Catalog number 292935000
Price (USD)
175.00
Each
-
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
175.00
Each
Add to cart
Chemical Identifiers
CAS14593-04-5
IUPAC Name3-amino-3-phenylpropan-1-ol
Molecular FormulaC9H13NO
InChI KeySEQXIQNPMQTBGN-UHFFFAOYNA-N
SMILESNC(CCO)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=92 %
Appearance (Form)Needles
Melting point70°C to 77°C
Appearance (Color)White
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0348017Certificate of AnalysisNov 26, 2021292935000, 292930025
1 result displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection