alpha-Terpineol, 97+%
alpha-Terpineol, 97+%
alpha-Terpineol, 97+%
alpha-Terpineol, 97+%
Thermo Scientific Chemicals

alpha-Terpineol, 97+%

CAS: 98-55-5 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Catalog number 301615000
Price (USD)
73.65
Special offer
Online exclusive
86.00
Save 12.35 (14%)
Each
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
86.00
Save 12.35 (14%)
Each
Add to cart
Chemical Identifiers
CAS98-55-5
IUPAC Name2-[(1R)-4-methylcyclohex-3-en-1-yl]propan-2-ol
Molecular FormulaC10H18O
InChI KeyWUOACPNHFRMFPN-VIFPVBQESA-N
SMILESCC1=CC[C@@H](CC1)C(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Viscous liquid or low melting crystalline mass or
Refractive index1.4800 to 1.4850 (20°C, 589 nm)
Appearance (Color)Clear colorless to white
GC>=97.0 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468022Certificate of AnalysisJan 31, 2025301610250, 301615000, 301610025
A0465523Certificate of AnalysisSep 10, 2024301610250, 301615000, 301610025
A0456421Certificate of AnalysisJun 14, 2024301610250, 301615000, 301610025
A0463271Certificate of AnalysisMay 31, 2024301610250, 301615000, 301610025
A0461541Certificate of AnalysisApr 05, 2024301610250, 301615000, 301610025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection