Thermo Scientific™

NeutrAvidin Protein, AP

Catalog number: 31002
Have Questions?
Thermo Scientific™

NeutrAvidin Protein, AP

Catalog number: 31002
Have Questions?
Catalog Number
31002
Unit Size
2 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
310022 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific™ alkaline phosphatase-conjugated NeutrAvidin Protein is a specially-prepared form of avidin that decreases background in biotin-binding.

Features include:
Near-neutral isoelectric point—pI = 6.3, more neutral than native avidin
Nearly devoid of glycosylation—decreased possibility of lectin binding compared to native avidin
No RYD recognition sequence—no known off-target binding domains like streptavidin
Affordable—significantly less expensive than streptavidin

NeutrAvidin Protein is deglycosylated native avidin from egg whites. Removal of the excess carbohydrate by an exclusive process yields a protein with a more neutral isoelectric point and less nonspecific binding properties. Conjugated NeutrAvidin Protein provides exceptional performance in western blot, ELISA, and IHC applications that require biotin-binding probes. Assay specificity, sensitivity, and signal-to-noise ratios with NeutrAvidin Protein are generally equivalent or better than with the significantly more expensive streptavidin.

Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles, and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin-binding proteins. The specific activity for biotin binding is approximately 14 μg/mg of protein, which is near the theoretical maximum activity.

Applications:
In situ hybridization—NeutrAvidin-AP Conjugate is currently being used by the Allen Institute for Brain Science to create the Allen Brain Atlas.
Western blotting—use NeutrAvidin Protein conjugates (HRP or AP) instead of streptavidin conjugates for detection of biotinylated target proteins or biotinylated primary or secondary antibodies in western blots.
ELISA—NeutrAvidin Protein conjugates (HRP or AP) replace corresponding streptavidin conjugates for direct or indirect sandwich ELISA detection schemes involving biotin-labeled proteins and antibodies. Depending on the specific assay components (e.g., blocking buffers, etc.), NeutrAvidin Protein conjugates provide similar or better assay performance measured as signal-to-noise ratio.

Related products
NeutrAvidin™ Protein
NeutrAvidin™ Protein, Horseradish Peroxidase Conjugated
NeutrAvidin™ Protein, Fluorescein Conjugated
NeutrAvidin™ Protein, Maleimide-Activated

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Conjugate
AP
Quantity
2 mg
Form
Solid
Protein Family
Avidin
Target Molecule
Biotin
Label or Dye
AP (Alkaline Phosphatase)

Contents & Storage

Upon receipt store product at –20°C. Product is shipped on dry ice.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text