2-(Trifluoromethyl)benzoyl chloride, 98%
2-(Trifluoromethyl)benzoyl chloride, 98%
2-(Trifluoromethyl)benzoyl chloride, 98%
2-(Trifluoromethyl)benzoyl chloride, 98%
Thermo Scientific Chemicals

2-(Trifluoromethyl)benzoyl chloride, 98%

CAS: 312-94-7 | C8H4ClF3O | 208.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 312930050
Price (EUR)
30,18
Online Exclusive
35,50
Save 5,32 (15%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
30,18
Online Exclusive
35,50
Save 5,32 (15%)
Each
Add to cart
Chemical Identifiers
CAS312-94-7
IUPAC Name2-(trifluoromethyl)benzoyl chloride
Molecular FormulaC8H4ClF3O
InChI KeyMXIUWSYTQJLIKE-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4780 to 1.4810 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear yellow to pink
GC>=97.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0305877Certificate of AnalysisOct 04, 2023312930010, 312930050, 312930250
A0400777Certificate of AnalysisSep 25, 2023312930010, 312930050, 312930250
A0437698Certificate of AnalysisJan 07, 2022312930010, 312930050, 312930250
A0436483Certificate of AnalysisDec 22, 2021312930010, 312930050, 312930250
4 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 1
Serious Eye Damage/Eye Irritation
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H314 - Causes severe skin burns and eye damage

EU Specific Hazard Statements

EUH029 - Contact with water liberates toxic gas

Precautionary Statements

P402 + P404 - Store in a dry place. Store in a closed container

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P302 + P350 - IF ON SKIN: Gently wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Share catalog number, name or link

1x1 image pixel for data collection