Thermo Scientific™

Pierce™ ECL Western Blotting Substrate

Catalog number: 32209
Have Questions?
Thermo Scientific™

Pierce™ ECL Western Blotting Substrate

Catalog number: 32209
Have Questions?
Catalog Number
32209
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 250 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
32209250 mL
-
3210950 mL
-
32209X44 x 250 mL
-
32106X44 x 500 mL
-
32106500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce ECL Western Blotting Substrate is a value-priced, entry-level horseradish peroxidase (HRP) substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.

Pierce ECL Plus Western Blotting Substrate characteristics include:
Sensitivity: low picogram
Stability: 1-hr working solution stability, 1-year kit stability at 4°C
Compatibility: nitrocellulose and PVDF membranes
Signal duration: 1 to 2 hours
Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–10 μg/mL)
Recommended secondary antibody concentration: 1:1,000–1:15,000 dilution (0.07–1.0 μg/mL)

Compare and learn more about our western blot chemiluminescent substrates ›
View all HRP substrates available ›

Pierce ECL Western Blotting Substrate provides reliability and performance equivalent to other standard ECL substrates for detection of HRP enzyme activity. It enables the detection of picogram amounts of antigen and allows for easy detection of HRP using photographic or other imaging methods.

Because the luminol and peroxide reagent formulations are identical to other commercially available substrate products, one can switch to Pierce ECL without needing to optimize probing conditions or incubation protocols. Pierce ECL Western Blotting Substrate provides performance identical to the original Amersham ECL Western Blotting Detection Reagent from GE Healthcare.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Substrate
Detection Method
Chemiluminescent
Form
Liquid
For Use With (Application)
Western Blot
Membrane Compatibility
Nitrocellulose, PVDF
Product Line
Pierce™
Quantity
250 mL
Substrate
Horseradish Peroxidase
Recommended Antibody Concentrations
2°: 1:1-15K (0.07-1μg/mL), 1°: 1:1K (0.2- 10μg/mL)
Sensitivity
Low Picogram
Signal Duration
1 to 2 hr.
Sufficient For
2500 sq. cm

Contents & Storage

Upon receipt store at 2-8°C. Product is shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text