1x1 image pixel for data collection
Pierce™ ECL Western Blotting Substrate
Pierce™ ECL Western Blotting Substrate
Thermo Scientific™

Pierce™ ECL Western Blotting Substrate

Thermo Scientific Pierce ECL Western Blotting Substrate is a value-priced, entry-level horseradish peroxidase (HRP) substrate for enhanced chemiluminescence (ECL) thatRead more
Promo ImagePromotion
Promo code:RBLOT25
Save up to 24% on Western Blot Reagents and Gels
From now until June 30th, get 16% off when you spend $400 or more, and get 24% off when you spend $750 or more on select western blot reagent and protein gels.Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
32106Promo Image500 mL
32109Promo Image50 mL
32209X44 x 250 mL
32209Promo Image250 mL
32106X44 x 500 mL
Catalog number 32106
Price (USD)
271.65
Online Exclusive
310.00
Save 38.35 (12%)
Each
In stock
Add to cart
Quantity:
500 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
271.65
Online Exclusive
310.00
Save 38.35 (12%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce ECL Western Blotting Substrate is a value-priced, entry-level horseradish peroxidase (HRP) substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.

Pierce ECL Plus Western Blotting Substrate characteristics include:
Sensitivity: low picogram
Stability: 1-hr working solution stability, 1-year kit stability at 4°C
Compatibility: nitrocellulose and PVDF membranes
Signal duration: 1 to 2 hours
Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–10 μg/mL)
Recommended secondary antibody concentration: 1:1,000–1:15,000 dilution (0.07–1.0 μg/mL)

Compare and learn more about our western blot chemiluminescent substrates ›
View all HRP substrates available ›

Pierce ECL Western Blotting Substrate provides reliability and performance equivalent to other standard ECL substrates for detection of HRP enzyme activity. It enables the detection of picogram amounts of antigen and allows for easy detection of HRP using photographic or other imaging methods.

Because the luminol and peroxide reagent formulations are identical to other commercially available substrate products, one can switch to Pierce ECL without needing to optimize probing conditions or incubation protocols. Pierce ECL Western Blotting Substrate provides performance identical to the original Amersham ECL Western Blotting Detection Reagent from GE Healthcare.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateHRP
DescriptionPierce ECL Western Blotting Substrate
Kit ContentsLuminol/Enhancer, 250mL, Stable Peroxide Buffer, 250mL
Recommended Antibody Concentrations2°: 1:1-15K (0.07-1μg/mL), 1°: 1:1K (0.2- 10μg/mL)
SensitivityLow Picogram
Signal Duration1 to 2 hr.
Sufficient For5000 sq. cm
Detection MethodChemiluminescence
For Use With (Application)Western Blot
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LinePierce™
Product TypeSubstrate
Quantity500 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
Upon receipt store at 2-8°C. Product is shipped at ambient temperature.
iBlot 3 Western Blot Transfer System

Introducing iBlot 3 Western Blot Transfer System

Featuring higher throughput and built-in cooling for consistent protein transfer
Learn more

Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

How much more sensitive is SuperSignal West Femto Maximum Sensitivity Substrate than the other enhanced chemiluminescent (ECL) substrates?

SuperSignal West Femto Maximum Sensitivity Substrate is about 25- to 50-fold more sensitive than GE Healthcare ECL Substrate or Thermo Scientific ECL Substrate (Cat. No. 32106). SuperSignal West Femto Maximum Sensitivity Substrate is also significantly more sensitive than available chemiluminescent substrates for alkaline phosphatase. Please also see Tech Tip # 32 Guide to Enzyme Substrates for Western Blotting (https://tools.thermofisher.com/content/sfs/brochures/TR0032-Substrates-Western.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How much more sensitive is SuperSignal West Dura Extended Duration Substrate than the other enhanced chemiluminescent (ECL) substrates?

SuperSignal West Dura Extended Duration Substrate is about 10-fold more sensitive than GE Healthcare ECL Substrate or Thermo Scientific ECL Substrate (Cat. No. 32106). SuperSignal West Dura Extended Duration Substrate is also significantly more sensitive than available chemiluminescent substrates for alkaline phosphatase. Please also see Tech Tip: Guide to Enzyme Substrates for Western Blotting (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0032-Substrates-Western.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How can I increase the sensitivity of my Western blot?

Consider transferring to a different membrane or using a different detection method. We have observed increased sensitivity when using PVDF membranes in place of nitrocellulose. On PVDF membranes, using as little as 1 µg of total rat brain protein, PKC can be detected with alkaline phosphatase-mediated chromogenic detection in some cases using affinity-purified antibodies at a concentration of 0.5 µg/mL. Detection sensitivity can also be increased by using chemiluminescent detection, especially when using a SuperSignal West enhanced chemiluminescence subtrate (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/detect-proteins-western-blot/western-blot-detection-reagents/detection-technologies-western-blotting/chemiluminescent-western-blot-detection/supersignal-chemiluminescent-substrates.html) such as SuperSignal West Pico PLUS, SuperSignal West Dura, or SuperSignal West Femto. The secondary antibody should be used as recommend by the manufacturer and optimized as needed.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3213865ACertificate of AnalysisJun 26, 202532209
3213865Certificate of AnalysisJun 26, 202532209
3213864ACertificate of AnalysisJun 19, 202532109
3213864Certificate of AnalysisJun 19, 202532109
3213985ACertificate of AnalysisJun 03, 202532106
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection