Low DNA Binding Snap Cap Microcentrifuge Tube
Low DNA Binding Snap Cap Microcentrifuge Tube
Thermo Scientific™
Low DNA Binding Snap Cap Microcentrifuge Tube
Microcentrifuge Tubes ensure optimal sample integrity with minimal DNA loss. These tubes provide secure closure and reliable performance, perfect for sensitive applications in molecular biology.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
3405-DLB2 mL
3403-DLB0.6 mL
3403-DLBPK0.6 mL
3404-DLB1.5 mL
3404-DLBPK1.5 mL
3405-DLBPK2 mL
Catalog number 3405-DLB
Price (USD)
357.00
Case of 5000
Estimated availability date  Pending  
Add to cart
Volume (Metric):
2 mL
Recurring order eligible. Learn more »
Price (USD)
357.00
Case of 5000
Add to cart
Ask our AI about this Product
The importance of low DNA binding microcentrifuge tubes cannot be overstated. These tubes are specifically engineered to minimize the adhesion of DNA to the tube walls, which is crucial in preserving the quantity and quality of DNA samples during various laboratory procedures.
Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results. Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.
Specifications
Volume (Metric)2 mL
AutoclavableAutoclavable
Bottom ShapeConical
Closure TypeSnap Cap
ColorNatural
Fit for PurposePCR Ready
GraduatedYes
MaterialPolypropylene Resin
Product TypeMicrocentrifuge Tube
ShapeCylindrical
SterilityNon-sterile
Unit SizeCase of 5000
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Share catalog number, name or link
1x1 image pixel for data collection