Thermo Scientific™

SuperSignal™ West Femto Maximum Sensitivity Substrate

Catalog number: 34095X4
Have Questions?
Thermo Scientific™

SuperSignal™ West Femto Maximum Sensitivity Substrate

Catalog number: 34095X4
Have Questions?
Catalog Number
34095X4
Unit Size
4 x 100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 200 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
34095X44 x 100 mL
-
3409420 mL
-
34095100 mL
-
34096X44 x 200 mL
-
34096200 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific SuperSignal West Femto Maximum Sensitivity Substrate is an ultra-sensitive enhanced chemiluminescent (ECL) substrate for low femtogram protein level detection by western blot analysis with horseradish peroxidase (HRP) enzyme.

SuperSignal West Femto Maximum Sensitivity Substrate characteristics include:
Sensitivity: low-femtogram (mid-zeptomole)
Stability: 8-hr working solution stability, 1-year kit stability at 4°C
Compatibility: nitrocellulose and PVDF membranes
Signal duration: up to 8 hours
Recommended primary antibody concentration: 1:5,000–1:100,000 dilution (10–200 ng/mL)
Recommended secondary antibody concentration: 1:100,000–1:500,000 dilution (2–10 ng/mL)

Compare and learn more about our western blot chemiluminescent substrates ›
View all HRP substrates available ›

SuperSignal West Femto Maximum Sensitivity Substrate is designed for low femtogram-level detection by western blot analysis. When combined with optimized antibody concentrations and blocking buffers, SuperSignal West Femto substrate enables detection of target proteins in amounts that are too small to be seen with typical ECL substrates.

Specifications

Product Type
Substrate
Detection Method
Chemiluminescent
Form
Liquid
Product Line
SuperSignal™
Quantity
4 x 100 mL
Substrate
Horseradish Peroxidase
Recommended Antibody Concentrations
2°: 1:100-500 K (2-10 ng/mL)
1°: 1:5 K (10- 200 ng/mL)
Sensitivity
Low to mid-femtogram
Signal Duration
6 to 8 hr.
Kit Contents
Luminol/Enhancer, 50 mL

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text