Thermo Scientific™
Snap Cap Low Retention Microcentrifuge Tubes
Catalog number: 3446
Have Questions?
Thermo Scientific™
Snap Cap Low Retention Microcentrifuge Tubes
Catalog number: 3446
Have Questions?
Improve sample recovery by significantly reducing surface binding with Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes. Viscous liquids and costly reagents retained in a microcentrifuge tube effect assay accuracy and precision. With no additives or coatings, these tubes — available in 0.6mL, 1.5mL and 2.0mL sizes — offer maximum sample recovery by significantly reducing surface binding.
Catalog Number
3446
Unit Size
Case of 10000
Description
0.6mL Graduated, Nonsterile
Color
Natural
Max. RCF
30,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Cap StyleFlat
Max. RCF30,000 x g
Certifications/ComplianceCertified free of RNase, DNase, DNA and pyrogens
Description0.6mL Graduated, Nonsterile
Capacity (Metric)0.6 mL
ColorNatural
MaterialPolypropylene
Packaging100 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilityNon-sterile
GraduatedYes
Unit SizeCase of 10000
Catalog Number
Unit Size
Case of 5000
Description
2.0mL Graduated, Sterile
Color
Natural
Max. RCF
25,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Cap StyleFlat
Max. RCF25,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description2.0mL Graduated, Sterile
Capacity (Metric)2 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizeCase of 5000
Catalog Number
Unit Size
Pack of 500
Description
2.0mL Graduated, Sterile
Color
Natural
Max. RCF
25,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Cap StyleFlat
Max. RCF25,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description2.0mL Graduated, Sterile
Capacity (Metric)2 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizePack of 500
Catalog Number
Unit Size
Case of 5000
Description
2.0mL Graduated, Nonsterile
Color
Natural
Max. RCF
25,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Cap StyleFlat
Max. RCF25,000 x g
Certifications/ComplianceCertified free of RNase, DNase, DNA and pyrogens
Description2.0mL Graduated, Nonsterile
Capacity (Metric)2 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilityNon-sterile
GraduatedYes
Unit SizeCase of 5000
Catalog Number
Unit Size
Pack of 500
Description
2.0mL Graduated, Nonsterile
Color
Natural
Max. RCF
25,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Cap StyleFlat
Max. RCF25,000 x g
Certifications/ComplianceCertified free of RNase, DNase, DNA and pyrogens
Description2.0mL Graduated, Nonsterile
Capacity (Metric)2 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilityNon-sterile
GraduatedYes
Unit SizePack of 500
Catalog Number
Unit Size
Case of 5000
Description
1.5mL Graduated, Sterile
Color
Natural
Max. RCF
26,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Cap StyleFlat
Max. RCF26,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description1.5mL Graduated, Sterile
Capacity (Metric)1.5 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizeCase of 5000
Catalog Number
Unit Size
Pack of 500
Description
1.5mL Graduated, Sterile
Color
Natural
Max. RCF
26,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Cap StyleFlat
Max. RCF26,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description1.5mL Graduated, Sterile
Capacity (Metric)1.5 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 1000
Description
0.6mL Graduated, Nonsterile
Color
Natural
Max. RCF
30,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Cap StyleFlat
Max. RCF30,000 x g
Certifications/ComplianceCertified free of RNase, DNase, DNA and pyrogens
Description0.6mL Graduated, Nonsterile
Capacity (Metric)0.6 mL
ColorNatural
MaterialPolypropylene
Packaging100 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilityNon-sterile
GraduatedYes
Unit SizePack of 1000
Catalog Number
Unit Size
Case of 10000
Description
0.6mL Graduated, Sterile
Color
Natural
Max. RCF
30,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Cap StyleFlat
Max. RCF30,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description0.6mL Graduated, Sterile
Capacity (Metric)0.6 mL
ColorNatural
MaterialPolypropylene
Packaging100 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizeCase of 10000
Catalog Number
3449PK
also known as 3449
Unit Size
Pack of 1000
Description
0.6mL Graduated, Sterile
Color
Natural
Max. RCF
30,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Cap StyleFlat
Max. RCF30,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description0.6mL Graduated, Sterile
Capacity (Metric)0.6 mL
ColorNatural
MaterialPolypropylene
Packaging100 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizePack of 1000
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeDescriptionColorMax. RCFPrice (USD)AvailabilityQuantity
3446Full specifications
Case of 100000.6mL Graduated, NonsterileNatural30,000 x g
-
Cap StyleFlat
Max. RCF30,000 x g
Certifications/ComplianceCertified free of RNase, DNase, DNA and pyrogens
Description0.6mL Graduated, Nonsterile
Capacity (Metric)0.6 mL
ColorNatural
MaterialPolypropylene
Packaging100 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilityNon-sterile
GraduatedYes
Unit SizeCase of 10000
3453Full specifications
Case of 50002.0mL Graduated, SterileNatural25,000 x g
-
Cap StyleFlat
Max. RCF25,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description2.0mL Graduated, Sterile
Capacity (Metric)2 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizeCase of 5000
3453PKFull specifications
Pack of 5002.0mL Graduated, SterileNatural25,000 x g
-
Cap StyleFlat
Max. RCF25,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description2.0mL Graduated, Sterile
Capacity (Metric)2 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizePack of 500
3434Full specifications
Case of 50002.0mL Graduated, NonsterileNatural25,000 x g
-
Cap StyleFlat
Max. RCF25,000 x g
Certifications/ComplianceCertified free of RNase, DNase, DNA and pyrogens
Description2.0mL Graduated, Nonsterile
Capacity (Metric)2 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilityNon-sterile
GraduatedYes
Unit SizeCase of 5000
3434PKFull specifications
Pack of 5002.0mL Graduated, NonsterileNatural25,000 x g
-
Cap StyleFlat
Max. RCF25,000 x g
Certifications/ComplianceCertified free of RNase, DNase, DNA and pyrogens
Description2.0mL Graduated, Nonsterile
Capacity (Metric)2 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilityNon-sterile
GraduatedYes
Unit SizePack of 500
3451Full specifications
Case of 50001.5mL Graduated, SterileNatural26,000 x g
-
Cap StyleFlat
Max. RCF26,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description1.5mL Graduated, Sterile
Capacity (Metric)1.5 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizeCase of 5000
3451PKFull specifications
Pack of 5001.5mL Graduated, SterileNatural26,000 x g
-
Cap StyleFlat
Max. RCF26,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description1.5mL Graduated, Sterile
Capacity (Metric)1.5 mL
ColorNatural
MaterialPolypropylene
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizePack of 500
3446PKFull specifications
Pack of 10000.6mL Graduated, NonsterileNatural30,000 x g
-
Cap StyleFlat
Max. RCF30,000 x g
Certifications/ComplianceCertified free of RNase, DNase, DNA and pyrogens
Description0.6mL Graduated, Nonsterile
Capacity (Metric)0.6 mL
ColorNatural
MaterialPolypropylene
Packaging100 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilityNon-sterile
GraduatedYes
Unit SizePack of 1000
3449Full specifications
Case of 100000.6mL Graduated, SterileNatural30,000 x g
-
Cap StyleFlat
Max. RCF30,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description0.6mL Graduated, Sterile
Capacity (Metric)0.6 mL
ColorNatural
MaterialPolypropylene
Packaging100 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizeCase of 10000
3449PK
also known as 3449
Full specifications
Pack of 10000.6mL Graduated, SterileNatural30,000 x g
-
Cap StyleFlat
Max. RCF30,000 x g
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Description0.6mL Graduated, Sterile
Capacity (Metric)0.6 mL
ColorNatural
MaterialPolypropylene
Packaging100 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilitySterile
GraduatedYes
Unit SizePack of 1000
Showing 10 of 10
  • The duckbill cap reduces the pressure required to open the tube with a wider contact surface, yet ensures the cap will remain closed during centrifugation
  • Low retention properties are not achieved by a coating or plastic-embedding processes, ensuring that samples will not be contaminated from these additives
  • Tubes will remain closed during centrifugation yet open easily with one hand as needed
  • Tubes are available in amber to protect your light-sensitive samples
  • Graduation marks on the microcentrifuge tube allow for effective sample reference
Color/Packaging  Options
  • A variety of packaging options are available to help with contamination control in the lab
  • For stringent sterility requirements, individually wrapped sterile tubes are available
  • Color options, in addition to natural and amber, are red, blue, green, yellow and violet and a mixed color packaging
Highest Quality
  • Manufactured from virgin polypropylene, these microcentrifuge tubes ensure sample integrity during boiling, freezing and centrifugation
  • Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
  • Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
Recommended for:
Daily laboratory usage including centrifugation and short-term sample storage