Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) horseradish peroxidase (HRP) substrate that enables picogram to high femtogram-level protein detection by western blot analysis.
SuperSignal West Pico PLUS Chemiluminescent Substrate characteristics include:
•
Sensitivity: pictogram to femtogram sensitivity
•
Stability: 8-hr working solution stability; 1-year kit stability at room temperature
•
Compatibility: nitrocellulose and PVDF membranes
•
Signal duration: 6 to 24 hours
•
Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–1.0 μg/mL)
•
Recommended secondary antibody concentration: 1:20,000–100,000 dilution (10–50 ng/mL)
Compare and learn more about our western blot chemiluminescent substrates ›View all HRP substrates available ›SuperSignal West Pico PLUS substrate is designed to provide excellent signal intensity and sensitivity for western blotting with HRP conjugates. The intensity of the light emission combined with the exceptional duration allows for the acquisition of multiple exposures to more easily obtain publication-quality blot images. SuperSignal West Pico PLUS substrate is compatible with different membranes, blocking reagents, and a wide range of antibody dilutions, making it an ideal choice for most western blotting applications.