Thermo Scientific™

SuperSignal™ West Pico PLUS Chemiluminescent Substrate

Catalog number: 34580X4
Have Questions?
Thermo Scientific™

SuperSignal™ West Pico PLUS Chemiluminescent Substrate

Catalog number: 34580X4
Have Questions?
Catalog Number
34580X4
Unit Size
4 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
34580X44 x 500 mL
-
3457920 mL
-
34577200 mL
-
345781 L
-
34580500 mL
-
34578X44 x 1 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) horseradish peroxidase (HRP) substrate that enables picogram to high femtogram-level protein detection by western blot analysis.

SuperSignal West Pico PLUS Chemiluminescent Substrate characteristics include:
Sensitivity: pictogram to femtogram sensitivity
Stability: 8-hr working solution stability; 1-year kit stability at room temperature
Compatibility: nitrocellulose and PVDF membranes
Signal duration: 6 to 24 hours
Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–1.0 μg/mL)
Recommended secondary antibody concentration: 1:20,000–100,000 dilution (10–50 ng/mL)

Compare and learn more about our western blot chemiluminescent substrates ›
View all HRP substrates available ›

SuperSignal West Pico PLUS substrate is designed to provide excellent signal intensity and sensitivity for western blotting with HRP conjugates. The intensity of the light emission combined with the exceptional duration allows for the acquisition of multiple exposures to more easily obtain publication-quality blot images. SuperSignal West Pico PLUS substrate is compatible with different membranes, blocking reagents, and a wide range of antibody dilutions, making it an ideal choice for most western blotting applications.

Specifications

Product Type
Substrate
Detection Method
Chemiluminescent
Form
Liquid
Product Line
SuperSignal™
Quantity
4 x 500 mL
Conjugate
HRP
Substrate
Horseradish Peroxidase
Recommended Antibody Concentrations
2°: 1:20-100 K (10-50 ng/mL)
1°: 1:1 K (0.2-1 μg/mL)
Sensitivity
Low picogram to high femtogram
Signal Duration
6 to 24 hr.
Kit Contents
SuperSignal West Pico PLUS Luminol/Enhancer, 4 x 500 mL
Shipping Condition
Room Temperature

Contents & Storage

Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text