Acriflavine hydrochloride
Acriflavine hydrochloride
Acriflavine hydrochloride
Acriflavine hydrochloride
Thermo Scientific Chemicals

Acriflavine hydrochloride

Acriflavine hydrochloride, C27H28Cl4N6, CAS Number-69235-50-3, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acrifiavine hydrochloride, unii-1s73vw819c, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride | CAS: 69235-50-3 | C27H27Cl3N6 | 541.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 346840100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS69235-50-3
IUPAC Name3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine dihydrochloride chloride
Molecular FormulaC27H27Cl3N6
InChI KeyMKLTXAHQKDVBLY-UHFFFAOYSA-N
SMILESCl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Loss on drying=<7 % (105°C, 1 h)
Appearance (Color)Orange-red to red-brown
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0465864Certificate of AnalysisSep 06, 2024346840100, 346841000, 346840250
A0463261Certificate of AnalysisMay 31, 2024346840100, 346841000, 346840250
A0458345Certificate of AnalysisDec 08, 2023346840100, 346841000, 346840250
A0457375Certificate of AnalysisNov 06, 2023346840100, 346841000, 346840250
A0371327Certificate of AnalysisDec 14, 2022346840100, 346841000, 346840250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection