Toluidine Blue O, pure
Toluidine Blue O, pure
Toluidine Blue O, pure
Toluidine Blue O, pure
Thermo Scientific Chemicals

Toluidine Blue O, pure

CAS: 92-31-9 | C15H16ClN3S | 305.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 348600250
Price (EUR)
118,15
Online Exclusive
139,00
Save 20,85 (15%)
Each
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
118,15
Online Exclusive
139,00
Save 20,85 (15%)
Each
Add to cart
Chemical Identifiers
CAS92-31-9
IUPAC Name3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
Molecular FormulaC15H16ClN3S
InChI KeyHNONEKILPDHFOL-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to black
Appearance (Form)Powder
Infrared spectrumConforms
UV spectrumComplies
NMRComplies
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470538Certificate of AnalysisMay 17, 2025348600050, 348600250, 348601000
A0465487Certificate of AnalysisOct 02, 2024348600050, 348600250, 348601000
A0462401Certificate of AnalysisJul 03, 2024348600050, 348600250, 348601000
A0460537Certificate of AnalysisFeb 28, 2024348600050, 348600250, 348601000
A0453660Certificate of AnalysisJan 22, 2024348600050, 348600250, 348601000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection