Thermo Scientific™
ART™ Non-Filtered Standard Hinged Rack Pipette Tips
Catalog number: 3552-HRPK
Have Questions?
Thermo Scientific™
ART™ Non-Filtered Standard Hinged Rack Pipette Tips
Catalog number: 3552-HRPK
Have Questions?
Access tips with one hand using Thermo ScientificTM ARTTM Nonfiltered Hinged Rack Pipette Tips, which is designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover--now you have a traditional lift-off when you need it.
Catalog Number
3552-HRPK
Unit Size
Pack of 960
Description
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)
200 μL
Sterility
Non-sterile
Fit for Purpose
Certified Quality
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)200 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)2 in.
Length (Metric)5.07cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizePack of 960
Catalog Number
Unit Size
Case of 3072
Description
ART Non-filtered, LR, Sterile, Hinged Rack
Volume (Metric)
1000 μL
Sterility
Sterile
Fit for Purpose
Sterile
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionART Non-filtered, LR, Sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilitySterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile
Unit SizeCase of 3072
Catalog Number
3101-05-HRPK
also known as 3101-05-HR
Unit Size
Pack of 768
Description
ART Non-filtered, LR, Sterile, Hinged Rack
Volume (Metric)
1000 μL
Sterility
Sterile
Fit for Purpose
Sterile
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionART Non-filtered, LR, Sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilitySterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile
Unit SizePack of 768
Catalog Number
Unit Size
Case of 3072
Description
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)
1000 μL
Sterility
Non-sterile
Fit for Purpose
Certified Quality
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizeCase of 3072
Catalog Number
3102-05-HRPK
also known as 3102-05-HR
Unit Size
Pack of 768
Description
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)
1000 μL
Sterility
Non-sterile
Fit for Purpose
Certified Quality
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizePack of 768
Catalog Number
Unit Size
Case of 3072
Description
ART Non-filtered, Non-Filtered, Sterile, Hinged Rack
Volume (Metric)
1000 μL
Sterility
Sterile
Fit for Purpose
Sterile
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionART Non-filtered, Non-Filtered, Sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilitySterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile
Unit SizeCase of 3072
Catalog Number
3101-HRPK
also known as 3101-HR
Unit Size
Pack of 768
Description
ART Non-filtered, Non-Filtered, Sterile, Hinged Rack
Volume (Metric)
1000 μL
Sterility
Sterile
Fit for Purpose
Sterile
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionART Non-filtered, Non-Filtered, Sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilitySterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile
Unit SizePack of 768
Catalog Number
Unit Size
Case of 3072
Description
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)
1000 μL
Sterility
Non-sterile
Fit for Purpose
Certified Quality
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizeCase of 3072
Catalog Number
Unit Size
Pack of 768
Description
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)
1000 μL
Sterility
Non-sterile
Fit for Purpose
Certified Quality
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizePack of 768
Catalog Number
Unit Size
Case of 4800
Description
ART Non-filtered, LR, Sterile, Hinged Rack
Volume (Metric)
300 μL
Sterility
Sterile
Fit for Purpose
Sterile
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionART Non-filtered, LR, Sterile, Hinged Rack
Volume (Metric)300 μL
FilteredNon-Filtered
SterilitySterile
ColorNatural
ModelART Non-filtered
Length (English)2.3 in.
Length (Metric)5.84cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile
Unit SizeCase of 4800
Showing 10 of 42Show more
Catalog NumberSpecificationsUnit SizeDescriptionVolume (Metric)SterilityFit for PurposePrice (USD)AvailabilityQuantity
3552-HRPKFull specifications
Pack of 960ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack200 μLNon-sterileCertified Quality
-
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)200 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)2 in.
Length (Metric)5.07cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizePack of 960
3101-05-HRFull specifications
Case of 3072ART Non-filtered, LR, Sterile, Hinged Rack1000 μLSterileSterile
-
DescriptionART Non-filtered, LR, Sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilitySterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile
Unit SizeCase of 3072
3101-05-HRPK
also known as 3101-05-HR
Full specifications
Pack of 768ART Non-filtered, LR, Sterile, Hinged Rack1000 μLSterileSterile
-
DescriptionART Non-filtered, LR, Sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilitySterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile
Unit SizePack of 768
3102-05-HRFull specifications
Case of 3072ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack1000 μLNon-sterileCertified Quality
-
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizeCase of 3072
3102-05-HRPK
also known as 3102-05-HR
Full specifications
Pack of 768ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack1000 μLNon-sterileCertified Quality
-
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizePack of 768
3101-HRFull specifications
Case of 3072ART Non-filtered, Non-Filtered, Sterile, Hinged Rack1000 μLSterileSterile
-
DescriptionART Non-filtered, Non-Filtered, Sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilitySterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile
Unit SizeCase of 3072
3101-HRPK
also known as 3101-HR
Full specifications
Pack of 768ART Non-filtered, Non-Filtered, Sterile, Hinged Rack1000 μLSterileSterile
-
DescriptionART Non-filtered, Non-Filtered, Sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilitySterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile
Unit SizePack of 768
3102-HRFull specifications
Case of 3072ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack1000 μLNon-sterileCertified Quality
-
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizeCase of 3072
3102-HRPKFull specifications
Pack of 768ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack1000 μLNon-sterileCertified Quality
-
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)3.5 in.
Length (Metric)8.89cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizePack of 768
3771-05-HRFull specifications
Case of 4800ART Non-filtered, LR, Sterile, Hinged Rack300 μLSterileSterile
-
DescriptionART Non-filtered, LR, Sterile, Hinged Rack
Volume (Metric)300 μL
FilteredNon-Filtered
SterilitySterile
ColorNatural
ModelART Non-filtered
Length (English)2.3 in.
Length (Metric)5.84cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile
Unit SizeCase of 4800
Showing 10 of 42Show more
Features:
  • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • Sterile tips are DNase, RNase, DNA, ATP, and Endotoxin Free
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout