1x1 image pixel for data collection
Brewers yeast
Thermo Scientific Chemicals

Brewers yeast

CAS: 68876-77-7
Have Questions?
Catalog NumberQuantity
3680800101 kg
Catalog number 368080010
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS68876-77-7
SpecificationsSpecification SheetSpecification Sheet
Microbial contamination=<700 /g (Yeast)
Microbial contamination(Salmonella) Absent in 375g
Microbial contamination=<10 /g (Escherichia coli)
Microbial contamination=<50000 /g (Total plate count)
Microbial contamination=<500 /g (Moulds)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0459854Certificate of AnalysisMay 17, 202510101152, 368080250, 368080050, 368080010
A0471146Certificate of AnalysisMay 17, 202510101152, 368080250, 368080050, 368080010
A0465448Certificate of AnalysisAug 26, 2024368080250, 368080050, 368080010
A0452537Certificate of AnalysisApr 25, 2024368080250, 368080050, 368080010
A0451667Certificate of AnalysisApr 02, 2024368080250, 368080050, 368080010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection