369678 Full specifications
Case of 24 Cryobank Universal Picker/Pen - Description Cryobank Universal Picker/Pen Capacity (Metric) 1 mL (Tube) Material Plastic Type Universal Picker/Pen Vial Type Cryobank, Bank-It Color Black No. per Pack 24 No. per Case 24 For Use With (Equipment) Cryobank and Bank-It Systems Unit Size Case of 24
374001 Full specifications
Case of 24 Cryobox divider, 13 x 13 grid - Description Cryobox divider, 13 x 13 grid Capacity (Metric) 0.5 mL (Tube), 1 mL (Tube) Holds 169 1 mL Nunc Cryobank Tubes Material Polycarbonate Type Cryobox Divider Vial Type Cryobank Color Black, White No. per Pack 24 No. per Case 24 Unit Size Case of 24
5050-0001 Full specifications
Case of 24 Polycarbonate CryoBox, Undivided - Description Polycarbonate CryoBox, Undivided Capacity (Metric) 1 mL (Tube) Material Polycarbonate Type Undivided CryoBox Coding 2D Barcode Vial Type Cryobank Color White No. per Pack 4 No. per Case 24 Unit Size Case of 24
5050-0001PK also known as 5050-0001
Full specifications
Pack of 4 Polycarbonate CryoBox, Undivided - Description Polycarbonate CryoBox, Undivided Capacity (Metric) 1 mL (Tube) Material Polycarbonate Type Undivided CryoBox Coding 2D Barcode Vial Type Cryobank Color White No. per Pack 4 No. per Case 24 Unit Size Pack of 4
5050-0169 Full specifications
Case of 24 Polycarbonate CryoBox with 13 x 13 divider - Description Polycarbonate CryoBox with 13 x 13 divider Capacity (Metric) 1 mL (Tube) Material Polycarbonate Type Cryobox Vial Type Cryobank Color White No. per Pack 4 No. per Case 24 For Use With (Equipment) Cryobank Unit Size Case of 24
5050-0169PK also known as 5050-0169
Full specifications
Pack of 4 Polycarbonate CryoBox with 13 x 13 divider - Description Polycarbonate CryoBox with 13 x 13 divider Capacity (Metric) 1 mL (Tube) Material Polycarbonate Type Cryobox Vial Type Cryobank Color White No. per Pack 4 No. per Case 24 For Use With (Equipment) Cryobank Unit Size Pack of 4
374115 Full specifications
Case of 845 1.0mL Nunc Cryobank Tubes, Blank, with Caps, Sterile, Loose Packed 1,316.70 Online price 1,386.00
- Description 1.0mL Nunc Cryobank Tubes, Blank, with Caps, Sterile, Loose Packed Capacity (Metric) 1.0 mL Holds 196 1 mL Nunc Cryobank Tubes Sterility Sterile Type Cryobank Tubes Coding None Vial Type Cryobank Color White, Clear (Closure) Packaging 169 tubes/bag No. per Pack 845 No. per Case 845 For Use With (Equipment) Cryobank Unit Size Case of 845