Double your cryobank storage capacity and cut storage costs by 50%. The Thermo Scientific™ Dense Storage Options use standard CryoBoxes™ with 13 x 13 tube separators and fit 169 1mL tubes in a single CryoBox™. These standard-sized boxes are compatible with existing standard-sized lab freezer racks.
Catalog Number
374115
Unit Size
Case of 845
Description
1.0mL Nunc Cryobank Tubes, Blank, with Caps, Sterile, Loose Packed
Price (USD)
1,316.70 Online price
1,386.00
Availability
-
Full specifications
Quantity
Description
1.0mL Nunc Cryobank Tubes, Blank, with Caps, Sterile, Loose Packed