Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Thermo Scientific Chemicals

Sand, pure, 40-100 mesh

CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 370942500
Price (EUR)
25,67
Online Exclusive
30,20
Save 4,53 (15%)
Each
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
25,67
Online Exclusive
30,20
Save 4,53 (15%)
Each
Add to cart
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Particle size207 µm (AGS) (typical)
Silicon dioxide (SiO2)99.77 % (typical)
Iron(III)oxide (Fe2O3)0.009 % (typical)
Aluminium oxide (Al2O3)0.030 % (typical)
Titanium dioxide (TiO2)0.019 % (typical)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473192Certificate of AnalysisJun 11, 2025370940250, 370940010, 370940050, 370942500
A0464720Certificate of AnalysisJul 31, 2024370940250, 370940010, 370940050, 370942500
A0426754Certificate of AnalysisApr 23, 2024370940250, 370940010, 370940050, 370942500
A0460096Certificate of AnalysisFeb 12, 2024370940250, 370940010, 370940050, 370942500
A0455771Certificate of AnalysisAug 25, 2023370940250, 370940010, 370940050, 370942500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection