Methyl acetate, 99+%, Extra Dry, AcroSeal™
Methyl acetate, 99+%, Extra Dry, AcroSeal™
Methyl acetate, 99+%, Extra Dry, AcroSeal™
Methyl acetate, 99+%, Extra Dry, AcroSeal™
Methyl acetate, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

Methyl acetate, 99+%, Extra Dry, AcroSeal™

CAS: 79-20-9 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 371831000
Price (USD)
44.65
Special offer
Online exclusive
Ends: 26-Sep-2025
52.50
Save 7.85 (15%)
Each
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 26-Sep-2025
52.50
Save 7.85 (15%)
Each
Add to cart
Chemical Identifiers
CAS79-20-9
IUPAC Namemethyl acetate
Molecular FormulaC3H6O2
InChI KeyKXKVLQRXCPHEJC-UHFFFAOYSA-N
SMILESCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Water=<0.005 %
GC>=99.0 %
Residue after evaporation=<0.002 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2543282Certificate of AnalysisJul 02, 2025371830025, 371831000, 371830010
2543281Certificate of AnalysisJul 02, 2025371830025, 371831000, 371830010
2542618Certificate of AnalysisJun 27, 2025371830025, 371831000, 371830010
2542617Certificate of AnalysisJun 27, 2025371830025, 371831000, 371830010
2529938Certificate of AnalysisMay 24, 2025371830025, 371831000, 371830010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection