Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | StartingBlock (TBS) Blocking Buffer, 100 mL |
Product Type | Blocking Buffer |
Quantity | 100 mL |
Form | Liquid |
For Use With (Application) | Western Blot, ELISA, Immunohistochemistry (IHC) |
Product Line | StartingBlock™ |
Formulation | Proprietary protein formulation in Tris-buffered saline at pH 7.5 |
Concentration | 1X |
Buffer | TBS |
Blocking Agent | Single purified protein |
Unit Size | 100 mL |
Description | StartingBlock (PBS) Blocking Buffer, 100 mL |
Product Type | Blocking Buffer |
Quantity | 100 mL |
Form | Liquid |
For Use With (Application) | Western Blot, ELISA, Immunohistochemistry (IHC) |
Product Line | StartingBlock™ |
Formulation | Proprietary protein formulation in phosphate-buffered saline, pH 7.5 |
Concentration | 1X |
Buffer | PBS |
Blocking Agent | Single purified protein |
Unit Size | 100 mL |
Description | StartingBlock (PBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | Western Blot, ELISA, Immunohistochemistry (IHC) |
Product Line | StartingBlock™ |
Formulation | Proprietary protein formulation in phosphate-buffered saline, pH 7.5 |
Concentration | 1X |
Buffer | PBS |
Blocking Agent | Single purified protein |
Unit Size | 1 L |
Description | StartingBlock (PBS) Blocking Buffer, 3 x 1 L |
Product Type | Blocking Buffer |
Quantity | 3 x 1 L |
Form | Liquid |
For Use With (Application) | Western Blot, ELISA, Immunohistochemistry (IHC) |
Product Line | StartingBlock™ |
Formulation | Proprietary protein formulation in phosphate-buffered saline, pH 7.5 |
Buffer | PBS |
Blocking Agent | Single purified protein |
Unit Size | 3 x 1 L |
Description | StartingBlock (TBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | Western Blot, ELISA, Immunohistochemistry (IHC) |
Product Line | StartingBlock™ |
Formulation | Proprietary protein formulation in phosphate-buffered saline, pH 7.5 |
Concentration | 1X |
Buffer | TBS |
Blocking Agent | Single purified protein |
Unit Size | 1 L |
Description | StartingBlock T20 (PBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | Western Blot, ELISA |
Product Line | StartingBlock™ |
Formulation | Proprietary protein formulation in phosphate-buffered saline, pH 7.5, with 0.05% Tween 20 |
Concentration | 1X |
Buffer | PBST |
Blocking Agent | Single purified protein |
Unit Size | 1 L |
Description | StartingBlock T20 (TBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | Western Blot, ELISA |
Product Line | StartingBlock™ |
Formulation | Proprietary protein formulation in phosphate-buffered saline, pH 7.5, with 0.05% Tween 20 |
Concentration | 1X |
Buffer | TBST |
Blocking Agent | Single purified protein |
Unit Size | 1 L |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
37579 | 100 mL | StartingBlock (TBS) Blocking Buffer, 100 mL | 51.65 Online price 55.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37578 | 100 mL | StartingBlock (PBS) Blocking Buffer, 100 mL | 51.65 Online price 55.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37538 | 1 L | StartingBlock (PBS) Blocking Buffer, 1 L | 233.65 Online price 253.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37538X3 | 3 x 1 L | StartingBlock (PBS) Blocking Buffer, 3 x 1 L | 567.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37542 | 1 L | StartingBlock (TBS) Blocking Buffer, 1 L | 235.65 Online price 254.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37539 | 1 L | StartingBlock T20 (PBS) Blocking Buffer, 1 L | 251.65 Online price 272.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37543 | 1 L | StartingBlock T20 (TBS) Blocking Buffer, 1 L | 248.65 Online price 270.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|