Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | PNPP Substrate Kit |
Product Type | ELISA Substrate |
Quantity | 1 Kit |
Detection Method | Colorimetric |
Product Line | Pierce™ |
Substrate Properties | Chemical Substrate |
Substrate Type | Alkaline Phosphatase |
Unit Size | 105 tablets |
Description | PNPP Powder |
Product Type | ELISA Substrate |
Quantity | 25 g |
Detection Method | Colorimetric |
Form | Powder |
Substrate Properties | Chemical Substrate |
Substrate Type | Alkaline Phosphatase |
Shipping Condition | Ambient |
Unit Size | 25 g |
Description | PNPP Tablets |
Product Type | ELISA Substrate |
Quantity | 105 Tablets |
Detection Method | Colorimetric |
Format | Tablet |
Substrate Properties | Chemical Substrate |
Substrate Type | Alkaline Phosphatase |
Shipping Condition | Wet Ice |
Unit Size | 105 tablets |
Description | 1-Step™ PNPP Substrate Solution |
Product Type | ELISA Substrate |
Quantity | 100 mL |
Detection Method | Colorimetric |
Product Line | 1-Step™ |
Substrate Properties | Chemical Substrate |
Substrate Type | Alkaline Phosphatase |
Shipping Condition | Ambient |
Unit Size | 100 mL |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
37620 | 105 tablets | PNPP Substrate Kit | 252.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
34045 | 25 g | PNPP Powder | 192.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
34047 | 105 tablets | PNPP Tablets | 198.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37621 | 100 mL | 1-Step™ PNPP Substrate Solution | 151.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific Pierce PNPP (p-Nitrophenyl Phosphate, Disodium Salt) is a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm.
Features of PNPP (p-nitrophenyl phosphate):
• One-component alkaline phosphatase (AP) substrate
• Absorbance maximum of 405 nm
• Develops a yellow hue in solution upon reaction with AP
• Safer to use—contains a non-mercury, non-azide preservative
The substrate is offered in several formats:
PNPP Substrate
• Powder Form
PNPP Substrate Tablets
• 5 mg tablets with an inert filler as the balance
PNPP Substrate Kit
• PNPP tablets and a Diethanolamine Buffer to yield more than one liter of substrate. The Diethanolamine Substrate Buffer (Cat. No. 34064) is also provided individually as a 5X concentrate.
1-Step™ PNPP Substrate Solution
• Circumvents this time-consuming and variable introducing step by providing a single component PNPP substrate. This substrate is stable for 12 months at 2–8°C and can be stopped with conventional methods.