PNPP Substrates
PNPP Substrates
Thermo Scientific™

PNPP Substrates

Thermo Scientific™ Pierce PNPP Substrates are powders, tablets, solutions and complete kits of p-nitrophenyl phosphate, a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormat
34045Powder
34047Tablet
37620Kit
37621Liqiuid
Catalog number 34045
Price (USD)
192.65
Online Exclusive
215.00
Save 22.35 (10%)
Each
In stock
Add to cart
Format:
Powder
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
192.65
Online Exclusive
215.00
Save 22.35 (10%)
Each
Add to cart
Ask our AI about this Product
PNPP (p-Nitrophenyl Phosphate, Disodium Sal) is a widely used substrate for detecting alkaline phosphatase in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm. Pierce PNPP is available in four formats: dry crystalline powder, single-use tablets, a stable substrate solution, and a complete kit containing substrate solution and diethanolamine buffer.

Thermo Scientific Pierce PNPP (p-Nitrophenyl Phosphate, Disodium Salt) is a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm.

Features of PNPP (p-nitrophenyl phosphate):
• One-component alkaline phosphatase (AP) substrate
• Absorbance maximum of 405 nm
• Develops a yellow hue in solution upon reaction with AP
• Safer to use—contains a non-mercury, non-azide preservative

The substrate is offered in several formats:
PNPP Substrate
• Powder Form

PNPP Substrate Tablets
• 5 mg tablets with an inert filler as the balance

PNPP Substrate Kit
• PNPP tablets and a Diethanolamine Buffer to yield more than one liter of substrate. The Diethanolamine Substrate Buffer (Cat. No. 34064) is also provided individually as a 5X concentrate.

1-Step™ PNPP Substrate Solution
• Circumvents this time-consuming and variable introducing step by providing a single component PNPP substrate. This substrate is stable for 12 months at 2–8°C and can be stopped with conventional methods.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPNPP Powder
Detection MethodColorimetric
FormPowder
FormatPowder
Shipping ConditionAmbient
Substrate PropertiesChemical Substrate
Substrate TypeAlkaline Phosphatase
Product TypeELISA Substrate
Quantity25 g
Unit SizeEach
Contents & Storage
Upon receipt store product at 4°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3216677Certificate of AnalysisJul 08, 202534047
79439789Certificate of AnalysisJun 25, 202537621
79439788Certificate of AnalysisJun 16, 202537621
79423016Certificate of AnalysisMay 16, 202537621
3212107Certificate of AnalysisMay 09, 202534047
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection