1x1 image pixel for data collection
Ammonium hydroxide, for analysis, 25% solution in water
Ammonium hydroxide, for analysis, 25% solution in water
Ammonium hydroxide, for analysis, 25% solution in water
Ammonium hydroxide, for analysis, 25% solution in water
Thermo Scientific Chemicals

Ammonium hydroxide, for analysis, 25% solution in water

CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 390030010
Price (USD)
64.65
Special offer
Online exclusive
75.80
Save 11.15 (15%)
Each
In stock
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
75.80
Save 11.15 (15%)
Each
Add to cart
Chemical Identifiers
CAS1336-21-6
IUPAC Nameamine hydrate
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILESN.O
View more
SpecificationsSpecification SheetSpecification Sheet
Bismuth (Bi)=<0.00001 %
Cobalt (Co)=<0.00001 %
Magnesium (Mg)=<0.00005 %
Sodium (Na)=<0.0001 %
Thallium (Tl)=<0.000005 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471174Certificate of AnalysisMay 17, 2025390030010, 390030025
A0471707Certificate of AnalysisMay 17, 2025390030010, 390030025
A0469043Certificate of AnalysisJan 31, 2025390030010, 390030025
A0468253Certificate of AnalysisDec 16, 2024390030010, 390030025
A0461904Certificate of AnalysisApr 23, 2024390030010, 390030025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection