Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set, 0.2 mL

Catalog number: 403086
Have Questions?
Applied Biosystems™

MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set, 0.2 mL

Catalog number: 403086
Have Questions?
Catalog Number
403086
Unit Size
100 assemblies
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 assemblies
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
403086100 assemblies
-
40308320 assemblies
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems™ MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set is a completely pre-assembled unit containing MicroAmp™ Reaction Tubes without caps already loaded in a MicroAmp™ 96-Well Tray/Retainer Set. MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Sets are optimally designed for precise PCR on Applied Biosystems™ 0.2 mL thermal cyclers.

• Completely pre-assembled for convenience
• Available in economical 100-unit bulk packs
• Non-sterile and autoclavable

Greater Sample Uniformity
Use with all instruments that have heated sample covers to eliminate condensation and the need for a mineral oil overlay. Proprietary MicroAmp™ Reaction Tubes are essential for high-performance PCR and ensure the greatest temperature and amplification uniformity for your samples. The MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set requires caps or full plate covers to seal the tubes.

Note: See user's manual or package insert for limited label license and trademark information.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
MicroAmp
Color
Natural
Product Type
96-Well Reaction Tube/Tray/Retainer Set
For Use With (Equipment)
2720 Thermal Cycler, 6100 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler
Plate Compatibility
96-Well Plates
Quantity
100 Assemblies

Contents & Storage

Contains 100 MicroAmp™ 96-Well Reaction Tube⁄Tray Retainer Sets. Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text